Skip to Content
Merck
All Photos(1)

Key Documents

33382

Supelco

Oxadiazon

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18Cl2N2O3
CAS Number:
Molecular Weight:
345.22
Beilstein:
558070
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)Oc1cc(N2N=C(OC2=O)C(C)(C)C)c(Cl)cc1Cl

InChI

1S/C15H18Cl2N2O3/c1-8(2)21-12-7-11(9(16)6-10(12)17)19-14(20)22-13(18-19)15(3,4)5/h6-8H,1-5H3

InChI key

CHNUNORXWHYHNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxadiazon is a herbicide, which is used for weed control management of obnoxious grasses and broad leaved weeds in a variety of crop plantations.

Application

Oxadiazon may be used as a reference standard in the determination of oxadiazon in soil, water and urine samples using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The degradation of oxadiazon and oxyfluorfen by photolysis.
Ying G-G and Williams B
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 34(4), 549-567 (1999)
Yang Zuo et al.
Bioorganic & medicinal chemistry, 20(1), 296-304 (2011-12-02)
Protoporphyrinogen oxidase (Protox, EC 1.3.3.4) has attracted great interest during the last decades due to its unique biochemical characteristics and biomedical significance. As a continuation of our research work on the development of new PPO inhibitors, 23 new 1,3,4-thiadiazol-2(3H)-ones bearing
Determination of tebufenpyrad and oxadiazon by solid-phase microextraction and gas chromatography-mass spectrometry.
Navalon A, et al.
Chromatographia, 54(5-6), 377-382 (2001)
Alberto Navalón et al.
Journal of chromatography. A, 946(1-2), 239-245 (2002-03-05)
A method for the determination of trace amounts of the herbicide oxadiazon was developed using headspace solid-phase microextraction (HS-SPME), gas chromatography-mass spectrometry (GC-MS) and selected ion monitoring. It was applied to determine oxadiazon in ground water, agricultural soil, must, wine
L Richert et al.
Toxicology and applied pharmacology, 141(1), 35-43 (1996-11-01)
Oxadiazon [5-ter-butyl-3-(2,4-dichloro-5-isopropoxyphenyl)- 1,3,4-oxadiazol-2(3H)-one] was administered orally at 20-500 mg/ kg body wt/day to male Sprague-Dawley CD rats for 14 days, at 20-200 mg/kg body wt/day to male CD1 [CR1/CD-1(1GR)BR] mice for 28 days, and at 500 mg/kg body wt/day to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service