Skip to Content
Merck
All Photos(1)

Key Documents

S356

Sigma-Aldrich

Salicylaldehyde

reagent grade, 98%

Synonym(s):

2-Hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein:
471388
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

4.2 (vs air)

vapor pressure

1 mmHg ( 33 °C)

Assay

98%

impurities

≤1% phenol

refractive index

n20/D 1.573 (lit.)

bp

197 °C (lit.)

mp

1-2 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1C=O

InChI

1S/C7H6O2/c8-5-6-3-1-2-4-7(6)9/h1-5,9H

InChI key

SMQUZDBALVYZAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Salicyladehyde (2-Hydroxybenzaldehyde) may be used in the preparation of pyrocatechol via Dakin reaction.[1]
Salicylaldehyde may be used as starting reagent in the synthesis of novel unsymmetrical chiral Salen Schiff base ligands.[2] It may be used in the preparation of salicylaldehyde phenylhydrazine, a useful indicator for the titration of organometallic reagents.[3]

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Use of Salicylaldehyde Phenylhydrazone as an Indicator for the Titration of Organometallic Reagents.
Brian E. Love et al.
The Journal of organic chemistry, 64(10), 3755-3756 (2001-10-25)
Eagleson M.
Concise Encyclopedia Chemistry, 301-301 (1994)
Salicin from host plant as precursor of salicylaldehyde in defensive secretion of chrysomeline larvae.
Pasteels JM, et al.
Physiological Entomology, 8(3), 307-314 (1983)
Unsymmetric chiral salen Schiff bases: A new chiral ligand pool from bis-schiff bases containing two different salicylaldehyde units.
Lopez J, et al.
Tetrahedron Letters, 39(24), 4199-4202 (1998)
Rhodium(III)-catalyzed dehydrogenative Heck reaction of salicylaldehydes.
Zhuangzhi Shi et al.
Angewandte Chemie (International ed. in English), 51(32), 8092-8096 (2012-06-30)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service