Merck
  • Novel D-xylose derivatives stimulate muscle glucose uptake by activating AMP-activated protein kinase alpha.
Novel D-xylose derivatives stimulate muscle glucose uptake by activating AMP-activated protein kinase alpha.
Journal of medicinal chemistry (2008-12-04)
Arie Gruzman, Ofer Shamni, Moriya Ben Yakir, Daphna Sandovski, Anna Elgart, Evgenia Alpert, Guy Cohen, Amnon Hoffman, Yehoshua Katzhendler, Erol Cerasi, Shlomo Sasson
ABSTRACT
Type 2 diabetes mellitus has reached epidemic proportions; therefore, the search for novel antihyperglycemic drugs is intense. We have discovered that D-xylose increases the rate of glucose transport in a non-insulin-dependent manner in rat and human myotubes in vitro. Due to the unfavorable pharmacokinetic properties of D-xylose we aimed at synthesizing active derivatives with improved parameters. Quantitative structure-activity relationship analysis identified critical hydroxyl groups in D-xylose. These data were used to synthesize various hydrophobic derivatives of D-xylose of which compound 19 the was most potent compound in stimulating the rate of hexose transport by increasing the abundance of glucose transporter-4 in the plasma membrane of myotubes. This effect resulted from the activation of AMP-activated protein kinase without recruiting the insulin transduction mechanism. These results show that lipophilic D-xylose derivatives may serve as prototype molecules for the development of novel antihyperglycemic drugs for the treatment of diabetes.
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
1,2-O-Isopropylidene-α-D-xylofuranose, 99%
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)