Merck
  • Proteogenomic Landscape of Breast Cancer Tumorigenesis and Targeted Therapy.
Proteogenomic Landscape of Breast Cancer Tumorigenesis and Targeted Therapy.
Cell (2020-11-20)
Karsten Krug, Eric J Jaehnig, Shankha Satpathy, Lili Blumenberg, Alla Karpova, Meenakshi Anurag, George Miles, Philipp Mertins, Yifat Geffen, Lauren C Tang, David I Heiman, Song Cao, Yosef E Maruvka, Jonathan T Lei, Chen Huang, Ramani B Kothadia, Antonio Colaprico, Chet Birger, Jarey Wang, Yongchao Dou, Bo Wen, Zhiao Shi, Yuxing Liao, Maciej Wiznerowicz, Matthew A Wyczalkowski, Xi Steven Chen, Jacob J Kennedy, Amanda G Paulovich, Mathangi Thiagarajan, Christopher R Kinsinger, Tara Hiltke, Emily S Boja, Mehdi Mesri, Ana I Robles, Henry Rodriguez, Thomas F Westbrook, Li Ding, Gad Getz, Karl R Clauser, David Fenyö, Kelly V Ruggles, Bing Zhang, D R Mani, Steven A Carr, Matthew J Ellis, Michael A Gillette
ABSTRACT
The integration of mass spectrometry-based proteomics with next-generation DNA and RNA sequencing profiles tumors more comprehensively. Here this "proteogenomics" approach was applied to 122 treatment-naive primary breast cancers accrued to preserve post-translational modifications, including protein phosphorylation and acetylation. Proteogenomics challenged standard breast cancer diagnoses, provided detailed analysis of the ERBB2 amplicon, defined tumor subsets that could benefit from immune checkpoint therapy, and allowed more accurate assessment of Rb status for prediction of CDK4/6 inhibitor responsiveness. Phosphoproteomics profiles uncovered novel associations between tumor suppressor loss and targetable kinases. Acetylproteome analysis highlighted acetylation on key nuclear proteins involved in the DNA damage response and revealed cross-talk between cytoplasmic and mitochondrial acetylation and metabolism. Our results underscore the potential of proteogenomics for clinical investigation of breast cancer through more accurate annotation of targetable pathways and biological features of this remarkably heterogeneous malignancy.
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Aprotinin Nicotiana tobacco, >= 5TIU/mg protein, >= 98 % SDS-PAGE | 9087-70-1, recombinant, expressed in Nicotiana (tobacco), ≥5 TIU/mg protein, ≥98% (SDS-PAGE)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%