Skip to Content
Merck
All Photos(1)

Key Documents

A2010

Sigma-Aldrich

Nα-Acetyl-L-lysine

Nα-Acetyl-L-lysine

≥98% (TLC), suitable for ligand binding assays

Synonym(s):

N2-acetyl-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

Nα-Acetyl-L-lysine,

Assay

≥98% (TLC)

Quality Level

form

powder

technique(s)

ligand binding assay: suitable

color

colorless to white

mp

256-258 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C8H16N2O3/c1-6(11)10-7(8(12)13)4-2-3-5-9/h7H,2-5,9H2,1H3,(H,10,11)(H,12,13)/t7-/m0/s1

InChI key

VEYYWZRYIYDQJM-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nα-Acetyl-L-lysine has been used as a substrate for lysyl oxidase for synthesis of authentic α-aminoadipic semialdehyde (AAS)-p-aminobenzoic acid (ABA).[1] It may be used in the synthesis of amino acid adducts of 4,4′-Methylenediphenyl diisocyanate (MDI).[2]

Biochem/physiol Actions

Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidase(s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Bi et al.
Emerging microbes & infections, 7(1), 108-108 (2018-06-15)
The DosR regulon is believed to be a key factor in latency adaptation of Mycobacterium tuberculosis and is strongly induced by multiple stresses, including hypoxia. Previous studies have revealed reversible acetylation of the conserved core DNA-binding lysine residue 182 (K182)
Wenhui Ren et al.
Scientific reports, 7, 39986-39986 (2017-01-04)
As members of bromodomain and extra-terminal motif protein family, bromodomain-containing proteins regulate a wide range of biological processes including protein scaffolding, mitosis, cell cycle progression and transcriptional regulation. The function of these bromodomain proteins (Brds) in innate immune response has
Yael Belo et al.
Biochimica et biophysica acta. General subjects, 1863(9), 1343-1350 (2019-06-07)
The signal transducer and activator of transcription 3 (STAT3) protein is activated by phosphorylation of a specific tyrosine residue (Tyr705) in response to various extracellular signals. STAT3 activity was also found to be regulated by acetylation of Lys685. However, the
Tenley C Archer et al.
Cancer cell, 34(3), 396-410 (2018-09-12)
There is a pressing need to identify therapeutic targets in tumors with low mutation rates such as the malignant pediatric brain tumor medulloblastoma. To address this challenge, we quantitatively profiled global proteomes and phospho-proteomes of 45 medulloblastoma samples. Integrated analyses
Sheng-Jun Fan et al.
Cell death & disease, 9(6), 655-655 (2018-05-31)
Alzheimer's disease (AD) is a dreadful neurodegenerative disease that leads to severe impairment of cognitive function, leading to a drastic decline in the quality of life. The primary pathological features of AD include senile plaques (SPs) and intracellular neurofibrillary tangles

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service