Skip to Content
Merck
All Photos(4)

Key Documents

302031

Sigma-Aldrich

Trifluoroacetic acid

suitable for HPLC, ≥99.0%

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.9 (vs air)

Quality Level

vapor pressure

97.5 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

technique(s)

HPLC: suitable
UV/Vis spectroscopy: suitable

impurities

<0.050% water

evapn. residue

<0.005%

refractive index

n20/D 1.3 (lit.)

pH

1 (20 °C, 10 g/L)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

solubility

water: soluble

density

1.489 g/mL at 20 °C (lit.)

λ

H2O reference

UV absorption

λ: 260 nm Amax: 0.9
λ: 270 nm Amax: 0.10
λ: 280 nm Amax: 0.05
λ: 290 nm Amax: 0.04
λ: 300 nm Amax: 0.03
λ: 320 nm Amax: 0.025

application(s)

food and beverages

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Appropriate for trace environmental analysis.
Used as a:
  • Mobile phase component in the estimation of chlorogenic acid and rutin in Rosaceae extracts by HPLC
  • Solvent to extract rabbit plasma, followed by HPLC purification of plasma protein content
  • Mobile phase solvent for separation of amino acids extracted from sediments, prior to HPLC analysis

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwang Su Kim et al.
ACS applied materials & interfaces, 5(9), 3591-3598 (2013-04-05)
Graphene-based electrochemical impedance sensors have recently received much attention due to their outstanding sensing capability and economic viability. In this study, we present a novel means of constructing an impedance sensing platform via harnessing intrinsic π-stacking interactions between probe protein
Continuous-flow synthesis of the anti-malaria drug artemisinin.
François Lévesque et al.
Angewandte Chemie (International ed. in English), 51(7), 1706-1709 (2012-01-18)
Luz M Hernández-Maldonado et al.
Nutrients, 11(7) (2019-07-25)
Mango (Mangifera indica L.) is a tropical fruit which is considered to be a source of dietary fiber (DF) and phenolic compounds (PCs). In this study, high DF mango-based fruit bars were developed from whole mango (peel and pulp). The
Guangfa Shi et al.
Organic letters, 17(1), 38-41 (2014-12-18)
Direct trifluoromethylation of arenes using TFA as the trifluoromethylating reagent was achieved with Ag as the catalyst. This reaction not only provides a new protocol for aryl C-H trifluoromethylation, but the generation of CF3· from TFA may prove useful in
Macro- and micromethods for the determination of serum vitamin A using trifluoroacetic acid.
J B Neeld et al.
The Journal of nutrition, 79, 454-462 (1963-04-01)

Articles

Optimization of a Reversed-Phase Liquid Chromatographic (RP-LC) method for the intact analysis of a therapeutic monoclonal antibody, trastuzumab, using a BIOshell™ A400 Protein C4 column.

Optimization of a Reversed-Phase Liquid Chromatographic (RP-LC) method for the intact analysis of a therapeutic monoclonal antibody, trastuzumab, using a BIOshell™ A400 Protein C4 column.

Optimization of a Reversed-Phase Liquid Chromatographic (RP-LC) method for the intact analysis of a therapeutic monoclonal antibody, trastuzumab, using a BIOshell™ A400 Protein C4 column.

Optimization of a Reversed-Phase Liquid Chromatographic (RP-LC) method for the intact analysis of a therapeutic monoclonal antibody, trastuzumab, using a BIOshell™ A400 Protein C4 column.

See All

Protocols

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service