Merck
  • Designed Parasite-Selective Rhomboid Inhibitors Block Invasion and Clear Blood-Stage Malaria.
Designed Parasite-Selective Rhomboid Inhibitors Block Invasion and Clear Blood-Stage Malaria.
Cell chemical biology (2020-09-06)
Shiv Gandhi, Rosanna P Baker, Sangwoo Cho, Stancho Stanchev, Kvido Strisovsky, Siniša Urban
ABSTRACT
Rhomboid intramembrane proteases regulate pathophysiological processes, but their targeting in a disease context has never been achieved. We decoded the atypical substrate specificity of malaria rhomboid PfROM4, but found, unexpectedly, that it results from "steric exclusion": PfROM4 and canonical rhomboid proteases cannot cleave each other's substrates due to reciprocal juxtamembrane steric clashes. Instead, we engineered an optimal sequence that enhanced proteolysis >10-fold, and solved high-resolution structures to discover that boronates enhance inhibition >100-fold. A peptide boronate modeled on our "super-substrate" carrying one "steric-excluding" residue inhibited PfROM4 but not human rhomboid proteolysis. We further screened a library to discover an orthogonal alpha-ketoamide that potently inhibited PfROM4 but not human rhomboid proteolysis. Despite the membrane-immersed target and rapid invasion, ultrastructural analysis revealed that single-dosing blood-stage malaria cultures blocked host-cell invasion and cleared parasitemia. These observations establish a strategy for designing parasite-selective rhomboid inhibitors and expose a druggable dependence on rhomboid proteolysis in non-motile parasites.
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Bis(trimethylsilyl)acetylene, 99%
Sigma-Aldrich
Benzyl isocyanide, 98%
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
4-Methoxyindole, 99%
Sigma-Aldrich
Batimastat, ≥98% (HPLC)
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Ubiquitin from bovine erythrocytes, BioUltra, ≥98% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Phosphocreatine disodium salt hydrate, ≥97%
Sigma-Aldrich
2-Nitrobenzyl alcohol, 97%
Supelco
Aloe-emodin, analytical standard
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Hemoglobin Assay Kit, sufficient for 250 colorimetric tests
Sigma-Aldrich
6-Nitroveratraldehyde, technical grade, 80%
Sigma-Aldrich
2-Amino-5-nitrobenzophenone, ≥98%
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)