Skip to Content
Merck
All Photos(1)

Documents

211737

Sigma-Aldrich

2-Amino-5-nitrobenzophenone

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(NO2)C(O)C6H5
CAS Number:
Molecular Weight:
242.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

166-168 °C (lit.)

SMILES string

Nc1ccc(cc1C(=O)c2ccccc2)[N+]([O-])=O

InChI

1S/C13H10N2O3/c14-12-7-6-10(15(17)18)8-11(12)13(16)9-4-2-1-3-5-9/h1-8H,14H2

InChI key

PZPZDEIASIKHPY-UHFFFAOYSA-N

General description

FT-IR and Raman spectra of 2-amino-5-nitrobenzophenone (ANBP) has been reported.

Application

2-Amino-5-nitrobenzophenone was used in the synthesis of [5-(4-nitrophenyl)-2-furyl]acrylic acid substituted benzophenone (anti-malarial agent).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Balachandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 118, 835-846 (2013-10-25)
The FT-IR and Raman spectra of 2-amino-5-nitrobenzophenone (ANBP) molecule have been recorded using Brucker IFS 66 V spectrometer in the range of 4000-100 cm(-1). The molecular geometry and vibrational frequencies in the ground state are calculated using the Hartree-Fock (HF)
Jochen Wiesner et al.
Bioorganic & medicinal chemistry letters, 13(3), 361-363 (2003-02-05)
We have developed the [5-(4-nitrophenyl)-2-furyl]acrylic acid substituted benzophenone 4g as a novel lead for anti-malarial agents. Here, we demonstrated that the acyl residue at the 2-amino group of the benzophenone core structure has to be a phenylacetic acid substructure substituted
Koichi Saito et al.
Chemical & pharmaceutical bulletin, 69(3), 258-264 (2021-03-02)
The degradation behavior of eight benzodiazepines (BZPs): alprazolam, etizolam, diazepam, triazolam, nitrazepam (NZP), flunitrazepam (FNZ), bromazepam, and lorazepam, in artificial gastric juice was monitored by a LC/photodiode array detector (PDA) to estimate their pharmacokinetics in the stomach. For drugs that
P J Cox et al.
International journal of pharmaceutics, 194(2), 147-153 (2000-02-29)
The role of single crystal diffraction in the quantitative determination of polymorphism is demonstrated by the examination of three compounds. Two polymorphs were found for each of the compounds bis(2-nitrophenyl) trisulphide (1), 2-amino-5-nitrobenzophenone (2) and bis(2-nitrophenyl) sulphide (3). Only in
T Inoue et al.
Journal of chromatography, 339(1), 163-169 (1985-04-12)
A method for the direct quantitative densitometry of nitrazepam and its main metabolites (7-aminonitrazepam, 7-acetamidonitrazepam and 2-amino-5-nitrobenzophenone) in urine was developed. The unchanged drug and its metabolites were extracted with benzene-dichloromethane (4:1), subjected to thin-layer chromatography, and determined by direct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service