Merck
  • Simple, sensitive and specific quantification of diamine oxidase activity in complex matrices using newly discovered fluorophores derived from natural substrates.
Simple, sensitive and specific quantification of diamine oxidase activity in complex matrices using newly discovered fluorophores derived from natural substrates.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2020-06-04)
Thomas Boehm, Matthias Karer, Elisabeth Gludovacz, Karin Petroczi, Marlene Resch, Kornelia Schuetzenberger, Kristaps Klavins, Nicole Borth, Bernd Jilma
ABSTRACT
To measure diamine oxidase (DAO) activity with high sensitivity in complex matrices like plasma or tissue extracts radioactive putrescine or horseradish peroxidase (HRP)/hydrogen peroxide (H2O2) coupling must be used. The use of radioactive material should be avoided and HRP/H2O2 coupling is compromised by antioxidants. Condensation of ortho-aminobenzaldehyde (oABA) with delta-1-pyrroline and delta-1-piperideine, the autocyclization products of the DAO-oxidized natural substrates putrescine and cadaverine, generates new quinazoline fluorophores with absorption and excitation maxima of 430 and 460 nm, respectively, and peak emission at 620 nm. Fluorescent-based detection limits are 20-40 times lower compared to absorption measurements. This assay can be used to measure DAO activity in human plasma after spiking recombinant human (rh)DAO, in rat plasma after intravenous rhDAO administration, in pregnancy plasma and in tissue extracts of DAO wild-type and knock-out mice. Using rat plasma the correlation between rhDAO activity and ELISA data is 99%. Human and rat plasma without DAO spiking and tissue extracts from DAO knock-out mice showed stable and low fluorescence in the presence of high substrate concentrations. Incubation of DAO with the natural substrates putrescine and cadaverine and oABA generates novel fluorophores increasing the detection limit compared to absorption measurements at least tenfold. This simple, sensitive and specific assay allows the non-radioactive quantification of DAO activity in complex matrices like plasma and tissue extracts without interference by antioxidants.
Sigma-Aldrich
2-Aminobenzaldehyde, ≥98%
Sigma-Aldrich
Diamine Oxidase from porcine kidney, ≥0.05 unit/mg solid
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
QuantiPro BCA Assay Kit, for 0.5-30 μg/ml protein
Sigma-Aldrich
Putrescine dihydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Corning® UV-Transparent Microplates, flat bottom clear, polystryrene, pkg of 50 ea, non-sterile, lid: no
Sigma-Aldrich
Cadaverine dihydrochloride, ~98%
Sigma-Aldrich
Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Diisopropyl ether, puriss. p.a., ≥98.5% (GC)