Skip to Content
Merck
All Photos(4)

Key Documents

D7876

Sigma-Aldrich

Diamine Oxidase from porcine kidney

≥0.05 unit/mg solid

Synonym(s):

Amine:oxygen oxidoreductase (deaminating) (pyridoxal-containing)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Porcine kidney

form

solid

specific activity

≥0.05 unit/mg solid

mol wt

170 kDa

solubility

100 mM sodium phosphate buffer, pH 7.2: soluble 10 mg/mL

foreign activity

monoamine oxidase (benzylamine substrate) ≤1%

storage temp.

−20°C

General description

Diamine oxidase from porcine kidney is a homodimer consisting of two equal subunits with a molecular weight of 87 kDa each. Each subunit contains one molecule of pyridoxal phosphate and one atom of copper. The molecular mass of the enzyme is found to be 170 kDa. The enzyme is a glycoprotein containing 5% hexose, 3.3% glucosamine, 2.6% N-acetylglucosamine, and 0.25% N-acetylneuraminic acid. The enzyme exhibits a high affinity for concanavalin A. Optimum pH with cadverine and histamine as substrates is found to be 6.3-7.4.

Application

Diamine Oxidase from porcine kidney has been used in the construction of histamine biosensor.
Diamine oxidase from porcine kidney has been used in a study to investigate a luminescence-based test for determining ornithine decarboxylase activity. Diamine oxidase from porcine kidney has also been used in a study to investigate N-linked oligosaccharide structures in diamine oxidase.

Biochem/physiol Actions

Diamine Oxidase catalyzes the oxidation of monoamines, diamines, and histamine to aldehydes, ammonia, and hydrogen peroxide. The enzyme is classified as a copper amine oxidase and it is a key enzyme in nitrogen metabolism. Diamine oxidase is inhibited by diethyldithiocarbamate, phenylhydrazine, semicarbazide, cyanide, isonicotinic acid hydrazide.

Unit Definition

One unit will oxidize 1.0 μmole of putrescine per hr at pH 7.2 at 37 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A PRELIMINARY INVESTIGATION ON A HISTAMINE BIOSENSOR CONSTRUCTED FROM DIAMINE OXIDASE IMMOBILISED ONTO AN OXYGEN PROBE
The Enzymes (1970)
Y Wang et al.
Analytical biochemistry, 287(2), 299-302 (2000-12-09)
A sensitive chemiluminescence-based method for the assay of ornithine decarboxylase (ODC) has been developed. This method, which permits the detection of putrescine (the product of ODC) at a picomolar range, can be used to determine ODC activity in cellular extracts.
HISTAMINE BIOSENSOR: A REVIEW
NorazlinaOthman F, et al.
The Malaysian Journal of Analytical Sciences (2006)
Taylor, H., and Tabor, C. W.
Methods in Enzymology, 17B, 735-740 null
A PRELIMINARY INVESTIGATION ON A HISTAMINE BIOSENSOR CONSTRUCTED FROM DIAMINE OXIDASE IMMOBILISED ONTO AN OXYGEN PROBE
Gupta N MM, et al.
International Journal of Pharmaceutical Sciences and Research (2018)

Protocols

To standardize a procedure for the enzymatic assay of Diamine Oxidase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service