Merck
  • Protein bioaccessibility from mycoprotein hyphal structure: In vitro investigation of underlying mechanisms.
Protein bioaccessibility from mycoprotein hyphal structure: In vitro investigation of underlying mechanisms.
Food chemistry (2020-06-26)
Raffaele Colosimo, Frederick J Warren, Tim J A Finnigan, Peter J Wilde
ABSTRACT
Mycoprotein is a food ingredient from filamentous fungi rich in protein and fibre. This study investigated the protein bioaccessibility from the fungal cells by colourimetric assays in different mycoprotein formulations, following extraction methods and in vitro gastrointestinal digestion. The methods effects were further analysed by static laser light scattering, SDS-PAGE and optical-fluorescence microscopy. The extraction methods released a comparable proportion of protein (30 wt%) independent of sample concentration (10 wt% and 25 wt%), whereas the simulated digestions endpoints released a higher proportion of protein from the less concentrated (46 wt%). Furthermore, mechanical/physical processing had only a minor impact. Intestinal proteases promoted the most efficient protein release but without causing any apparent damage to the cell walls when viewed by microscopy. This suggested that the enzymes can diffuse through the cell walls, due to its porosity/permeability, and are the main factors responsible for the hydrolysis and bioaccessibility of protein from mycoprotein.
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY)), for microscopy (Hist., Vit.)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Driselase from Basidiomycetes sp., powder, Protein ≥10 % by biuret
Solid-glass beads, borosilicate, diam. 1 mm
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Protein standard, Micro Standard, liquid
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Lactophenol blue solution, for microscopy, for staining molds
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Calcofluor White Stain, suitable for microbiology