Skip to Content
Merck
All Photos(1)

Key Documents

Z273619

Solid-glass beads

borosilicate, diam. 1 mm

Synonym(s):

borosilicate beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102422
NACRES:
NB.43

material

borosilicate

packaging

pack of 1 ea

diam.

1 mm

Looking for similar products? Visit Product Comparison Guide

General description

For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Soto et al.
Microbiology (Reading, England), 165(2), 174-187 (2019-01-17)
For micro-organisms cycling between free-living and host-associated stages, where reproduction occurs in both of these lifestyles, an interesting inquiry is whether evolution during the free-living stage can be positively pleiotropic to microbial fitness in a host environment. To address this
Pillai V Saranyan et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 206, 95-104 (2017-01-17)
This research investigated how ploidy level (diploid versus triploid) affects the heat shock protein (HSP) response in erythrocytes under different thermal stress regimes, both in vivo and in vitro, in Atlantic salmon (Salmo salar) and brook charr (Salvelinus fontinalis) in
Raffaele Colosimo et al.
Food chemistry, 330, 127252-127252 (2020-06-26)
Mycoprotein is a food ingredient from filamentous fungi rich in protein and fibre. This study investigated the protein bioaccessibility from the fungal cells by colourimetric assays in different mycoprotein formulations, following extraction methods and in vitro gastrointestinal digestion. The methods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service