Merck
  • Ecological and Phylogenetic Relationships Shape the Peripheral Olfactory Systems of Highly Specialized Gall Midges (Cecidomiiydae).
Ecological and Phylogenetic Relationships Shape the Peripheral Olfactory Systems of Highly Specialized Gall Midges (Cecidomiiydae).
Frontiers in physiology (2018-04-19)
Béla P Molnár, Tina Boddum, Sharon R Hill, Bill S Hansson, Ylva Hillbur, Göran Birgersson
ABSTRACT
Insects use sensitive olfactory systems to detect relevant host volatiles and avoid unsuitable hosts in a complex environmental odor landscape. Insects with short lifespans, such as gall midges (Diptera: Cecidomyiidae), are under strong selection pressure to detect and locate suitable hosts for their offspring in a short period of time. Ephemeral gall midges constitute excellent models for investigating the role of olfaction in host choice, host shift, and speciation. Midges mate near their site of emergence and females migrate in order to locate hosts for oviposition, thus females are expected to be more responsive to olfactory cues emitted by the host compared to males. In this study, we explored the correlation between host choice and the function of the peripheral olfactory system in 12 species of gall midges, including species with close phylogenetic relationships that use widely different host plants and more distantly related gall midge species that use similar hosts. We tested the antennal responses of males and females of the 12 species to a blend of 45 known insect attractants using coupled gas chromatographic-electroantennographic detection. When the species-specific response profiles of the gall midges were compared to a newly generated molecular-based phylogeny, we found they responded to the compounds in a sex- and species-specific manner. We found the physiological response profiles of species that use annual host plants, and thus have to locate their host every season, are similar for species with similar hosts despite large phylogenetic distances. In addition, we found closely related species with perennial hosts demonstrated odor response profiles that were consistent with their phylogenetic history. The ecology of the gall midges affects the tuning of the peripheral olfactory system, which in turn demonstrates a correlation between olfaction and speciation in the context of host use.
Sigma-Aldrich
Ethyl propionate, 99%
Sigma-Aldrich
3-(Methylthio)propyl isothiocyanate, ≥98%, FG
Sigma-Aldrich
Myrcene, stabilized, FCC, FG
Sigma-Aldrich
cis-3-Hexenyl benzoate, ≥97%, stabilized, FG
Sigma-Aldrich
cis-3-Hexenal solution, 50% in triacetin, stabilized
Sigma-Aldrich
trans-2-Hexen-1-al, 98%
Sigma-Aldrich
cis-2-Hexen-1-ol, 95%
Sigma-Aldrich
Hexyl acetate, 99%
Sigma-Aldrich
Isobutyl isobutyrate, ≥98%
Sigma-Aldrich
Camphene, 95%