Merck
  • High-performance liquid chromatographic method with coulometric detection for the determination of buspirone in human plasma by means of a column-switching technique.
High-performance liquid chromatographic method with coulometric detection for the determination of buspirone in human plasma by means of a column-switching technique.
Journal of chromatography. A (1998-05-16)
K Ary, K Róna, S Ondi, B Gachályi
ABSTRACT
A reversed-phase high-performance liquid chromatographic method with electrochemical detection has been developed for the determination of buspirone from human plasma. The separation was carried out by using a Supelcosil ABZ+ plus C18 reversed-phase column and 0.05 M potassium dihydrogenphosphate (pH 6.5)-acetonitrile (7:3, v/v) as the mobile phase. The compounds were detected by coulometry. Buspirone and the internal standard were extracted from the human plasma using Bond-Elut C18 solid-phase extraction cartridges. Following removal of the the highly lipophilic plasma components we applied a column-switching technique which reduced the duration of HPLC measurement from 60 min to 15 min. The limit of quantitation was found to be 100 pg/ml plasma.
Product Description
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL ABZ+Plus Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL ABZ+Plus Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
SUPELCOSIL ABZ+Plus Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL ABZ+Plus HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm