Skip to Content
Merck
All Photos(1)

Documents

59605

Supelco

SUPELCOSIL ABZ+Plus (5 µm) HPLC Columns

L × I.D. 2 cm × 2.1 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL ABZ+Plus Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm

Agency

suitable for USP L60

packaging

pkg of 2 ea

technique(s)

HPLC: suitable

L × I.D.

2 cm × 2.1 mm

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongping John Lin et al.
Journal of pharmaceutical and biomedical analysis, 37(4), 757-762 (2005-03-31)
A highly sensitive method was developed and validated for determining the free fraction of flunarizine in human plasma. Equilibrium dialysis was used for the separation of free (unbound) drug and liquid chromatography/tandem mass spectrometry (LC-MS/MS) was used for quantitation. Post-dialysis
Tânia M G Almeida et al.
Chemistry & biodiversity, 2(12), 1691-1700 (2006-12-29)
Absorption, distribution, metabolism, and excretion (ADME) properties are of invaluable importance to a bioactive compound. Permeation process is one of the most widely studied by many different techniques. Among them, reversed-phase liquid chromatography (RP-LC) is proving to be of great
K Ary et al.
Journal of chromatography. A, 797(1-2), 221-226 (1998-05-16)
A reversed-phase high-performance liquid chromatographic method with electrochemical detection has been developed for the determination of buspirone from human plasma. The separation was carried out by using a Supelcosil ABZ+ plus C18 reversed-phase column and 0.05 M potassium dihydrogenphosphate (pH
W Pan et al.
Journal of chromatography. B, Biomedical sciences and applications, 703(1-2), 129-138 (1998-02-03)
A sensitive, specific and precise HPLC-UV assay was developed to quantitate cocaine (COC) and its metabolites benzoylecgonine (BE), norcocaine (NC) and cocaethylene (CE) in rat plasma. After adding 50 microl of the internal standard solution (bupivacaine, 8 microg/ml) and 500
P Chaimbault et al.
Journal of chromatography. A, 855(1), 191-202 (1999-10-09)
A qualitative determination of 20 underivatized proteinic amino acids by LC-MS is reported. The need for chromatographic separation before mass spectrometry determination is demonstrated based on the study of several amino acid pairs which have some similar characteristics. Two suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service