59516
SUPELCOSIL™ LC-NH₂ NP (5 µm) HPLC Columns
L × I.D. 2 cm × 4 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
eCl@ss:
32110501
Recommended Products
Product Name
SUPELCOSIL™ LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Agency
suitable for USP L8
Quality Level
packaging
pkg of 2 ea
technique(s)
HPLC: suitable
L × I.D.
2 cm × 4 mm
matrix active group
amino phase
particle size
5 μm
application(s)
food and beverages
separation technique
normal phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Vijay V Upreti et al.
Biomedical chromatography : BMC, 17(6), 385-390 (2003-09-19)
A simple and sensitive high-performance liquid chromatography (HPLC) method has been developed and validated for the determination of DRF-1042, a novel orally active camptothecin (CPT) analog, in human plasma. The sample preparation was a simple deproteinization with acidified methanol yielding
S Djabarouti et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 799(1), 165-172 (2003-12-09)
The aim of this study was to develop a specific and sensitive high-performance liquid chromatographic (HPLC) assay for the determination of levofloxacin in human plasma, bronchoalveolar lavage and bone tissues. The sample extraction was based on a fully automated liquid-solid
E M Koves
Journal of chromatography. A, 692(1-2), 103-119 (1995-02-10)
A comprehensive approach to the analysis for many drugs in postmortem blood and biological fluids using high-performance liquid chromatography and diode array detection has been developed. To reduce the likelihood of co-eluting interference components of postmortem blood or other drugs
Kornélia Tekes
Journal of chromatographic science, 46(2), 169-173 (2008-03-28)
A sensitive, simple, and reliable high-performance liquid chromatographic method with electrochemical detection is developed for the measurement of four natural products, the serotonin-related indols from human platelet-rich plasma (PRP) using N-methylserotonin as internal standard. Separation of serotonin (5HT), 5-hydroxytryptophan (5HTP)
Sainath Babu et al.
Toxicology mechanisms and methods, 25(5), 410-416 (2015-05-30)
We have developed a simple, reversed-phase high-performance liquid chromatography (RP-HPLC) method for the determination of bisphenol A (BPA) in thermal paper cash register receipts (CRs). The method is suitable for analysis of other types of bisphenols and it involves an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service