Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium fluoride
Synonym(s):
Sodium fluoride
Linear Formula:
NaF
CAS Number:
7681-49-4
Molecular Weight:
41.99
EC Number:
231-667-8
Products
Product Category
salts (8)
Brand
SAFC (1)
Sigma-Aldrich (7)
Supelco (2)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Special Grade
ACS reagent (1)
Anhydrous (1)
Puriss (1)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
201154
Sodium fluoride,
ACS reagent, ≥99%
Expand
View Pricing
1.06449
Sodium fluoride,
for analysis EMSURE
®
Reag. Ph Eur
Expand
View Pricing
S1504
Sodium fluoride,
ReagentPlus
®
, ≥99%
Expand
View Pricing
215309
Sodium fluoride,
99.99% trace metals basis
Expand
View Pricing
1614002
Sodium fluoride,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
450022
Sodium fluoride,
anhydrous, powder, 99.99% trace metals basis
Expand
View Pricing
01148
Sodium fluoride,
puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Expand
View Pricing
S6776
Sodium fluoride,
BioReagent, suitable for insect cell culture, ≥99%
Expand
View Pricing
1.06450
Sodium fluoride,
99.99 Suprapur
®
Expand
View Pricing
S7920
Sodium fluoride,
BioXtra, ≥99%
Expand
View Pricing
1.72576
Sodium fluoride,
extra pure, for table water
Expand
View Pricing
PHR1408
Sodium fluoride,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing