Skip to Content
Merck
All Photos(1)

Key Documents

1359903

USP

Levocarnitine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

L-Carnitine inner salt, (−)-(R)-3-Hydroxy-4-(trimethylammonio)butyrate, Vitamin BT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2CH(OH)CH2COO-
CAS Number:
Molecular Weight:
161.20
Beilstein:
4292315
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levocarnitine

manufacturer/tradename

USP

mp

197-212 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[N+](C)(C)C[C@H](O)CC([O-])=O

InChI

1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m1/s1

InChI key

PHIQHXFUZVPYII-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levocarnitine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Levocarnitine Tablets
  • Levocarnitine Injection
  • Levocarnitine Oral Solution

Biochem/physiol Actions

Carnitine is a quaternary amine that occurs naturally in most mammalian tissue. It is present in relatively high concentrations in skeletal muscle and heart where it is involved in regulating energy metabolism. It shifts glucose metabolism from glycolysis to glycogen storage and enhances the transport of long chain fatty acids into the mitochondria where they are oxidized for energy production.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P D Pion et al.
The Veterinary clinics of North America. Small animal practice, 28(6), 1495-1514 (1999-03-31)
In this article, the current state of knowledge of the potential role of taurine and L-carnitine deficiency and supplementation in the genesis and management of cardiovascular disease in dogs is presented. There is limited direct evidence to support the recommendations
Yasuro Furuichi et al.
Biochimica et biophysica acta, 1837(10), 1699-1706 (2014-06-03)
Carnitine is well recognized as a key regulator of long-chain fatty acyl group translocation into the mitochondria. In addition, carnitine, as acetylcarnitine, acts as an acceptor of excess acetyl-CoA, a potent inhibitor of pyruvate dehydrogenase. Here, we provide a new
Reese H Clark et al.
Pediatrics, 134(1), e37-e46 (2014-06-11)
To describe the influence that gestational age and chronological age have on amino acid and acylcarnitine profiles in an at-risk population of premature infants. Metabolic profiles (15 amino acids and 35 acylcarnitines) were obtained by using standard newborn techniques on
Marieke G Schooneman et al.
Biochimica et biophysica acta, 1841(7), 987-994 (2014-04-22)
Acylcarnitines have been linked to obesity-induced insulin resistance. However the majority of these studies have focused on acylcarnitines in plasma. It is currently unclear to what extent plasma levels of acylcarnitines reflect tissue acylcarnitine metabolism. We investigated the correlation of
Allan Evans
American journal of kidney diseases : the official journal of the National Kidney Foundation, 41(4 Suppl 4), S13-S26 (2003-05-17)
Among the homeostatic processes controlling the endogenous L-carnitine pool in humans, the kidney has a vital role through extensive and adaptive tubular reabsorption. Kidney disease can lead to disturbances in L-carnitine homeostasis, and long-term hemodialysis therapy can lead to a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service