Skip to Content
Merck
All Photos(1)

Key Documents

1468501

USP

Norepinephrine bitartrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, L-(−)-Noradrenaline (+)-bitartrate salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

norepinephrine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.NC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C8H11NO3.C4H6O6.H2O/c9-4-8(12)5-1-2-6(10)7(11)3-5;5-1(3(7)8)2(6)4(9)10;/h1-3,8,10-12H,4,9H2;1-2,5-6H,(H,7,8)(H,9,10);1H2/t8-;1-,2-;/m01./s1

InChI key

LNBCGLZYLJMGKP-LUDZCAPTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

It is intended to be used as a USP reference standard in specified quality tests and assays.
Also used to prepare standard and system suitability solutions as described in the following United States Pharmacopeia (USP) monographs:
  • Norepinephrine Bitartrate
  • Norepinephrine Bitartrate Injection
  • Epinephrine
  • Racepinephrine
  • Epinephrine Bitartrate
  • Articaine Hydrochloride and Epinephrine Injection
  • Racepinephrine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norepinephrine Bitartrate Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 3198-3198 (2018)
Epinephrine Bitartrate
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(5), 1650-1650 (2021)
Articaine Hydrochloride and Epinephrine Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(3), 381-381 (2018)
Epinephrine
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(6), 1645-1645 (2020)
Racepinephrine
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(4), 3824-3824 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service