Skip to Content
Merck
All Photos(1)

Key Documents

Y0000882

Adrenaline

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Epinephrine, (−)-Adrenalin, (R)-(−)-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol, L-Adrenaline, L-Epinephrine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH(OH)CH2NHCH3
CAS Number:
Molecular Weight:
183.20
Beilstein:
2368277
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

adrenaline

manufacturer/tradename

EDQM

mp

215 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CNC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/t9-/m0/s1

InChI key

UCTWMZQNUQWSLP-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Adrenaline EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Adrenoceptor agonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Luongo et al.
European journal of oral implantology, 7(2), 187-199 (2014-07-01)
The aim of this prospective multicentre study was to evaluate the clinical outcome of immediately loaded single implants. Patients were recruited at six clinical centres. Inclusion criteria were singletooth replacement in fully healed sites or post-extraction sockets with adequate bone
M D De Bellis et al.
Biological psychiatry, 45(10), 1259-1270 (1999-06-01)
This investigation examined the relationship between trauma, psychiatric symptoms and urinary free cortisol (UFC) and catecholamine (epinephrine [EPI], norepinephrine [NE], dopamine [DA]) excretion in prepubertal children with posttraumatic stress disorder (PTSD) secondary to past child maltreatment experiences (n = 18)
Mads K Dalsgaard et al.
Experimental physiology, 89(3), 271-277 (2004-05-05)
Strenuous exercise increases the cerebral uptake of carbohydrate out of proportion to that of oxygen, but it is unknown whether such enhanced carbohydrate uptake is influenced by the marked endocrine response to exercise. During exhaustive exercise this study evaluated the
Candice Bjornson et al.
The Cochrane database of systematic reviews, (2)(2), CD006619-CD006619 (2011-02-18)
Croup is a common childhood illness characterized by barky cough, stridor, hoarseness and respiratory distress. Children with severe croup are at risk for intubation. Nebulized epinephrine (NE) may prevent intubation. To evaluate the efficacy and safety of NE in children
Phillip L Lieberman
The American journal of medicine, 127(1 Suppl), S6-11 (2014-01-05)
The International Classification of Diseases (ICD)-9 included a code only for anaphylactic shock. The improved ICD-10 coding defines the different symptoms and types of anaphylaxis and includes coding for anaphylaxis without shock. This new coding is consistent with the efforts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service