Skip to Content
Merck
All Photos(1)

Key Documents

1370270

USP

Loratadine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene-1-piperidinecarboxylic acid ethyl ester, Loratidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N2O2Cl
CAS Number:
Molecular Weight:
382.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

loratadine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(=O)N1CC\C(CC1)=C2/c3ccc(Cl)cc3CCc4cccnc24

InChI

1S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3

InChI key

JCCNYMKQOSZNPW-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Loratadine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Loratadine Capsules
  • Loratadine Chewable Tablets
  • Loratadine Oral Solution
  • Loratadine Orally Disintegrating Tablets
  • Loratadine Tablets

Biochem/physiol Actions

Non-sedating histamine H1-receptor antagonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I J Roman et al.
Clinical reviews in allergy, 11(1), 89-110 (1993-01-01)
Antihistamines are considered first-line therapy for the relief of symptoms from allergic rhinitis and chronic urticaria. The newer, second-generation, nonsedating antihistamines reduce the central nervous system and anticholinergic side effects commonly found with previous drugs. The availability of H1-receptor antagonists
M Haria et al.
Drugs, 48(4), 617-637 (1994-10-01)
Loratadine is a long-acting antihistamine agent, exhibiting partial selectivity for peripheral histamine H1-receptors. To date, loratadine has been evaluated in allergic rhinitis, urticaria and, to a limited extent, in asthma. In several large controlled comparative clinical studies, loratadine was superior
M R Namazi
Journal of the European Academy of Dermatology and Venereology : JEADV, 19(3), 319-322 (2005-04-29)
As the current antipsoriatic medications are commonly associated with deleterious side-effects, a determined search for safer agents, which could be used alone or in combination with current antipsoriatic drugs, would be very imperative. Psoriasis is believed to be characterized by
Safety and efficacy of loratadine in urticaria.
E W Monroe
International journal of dermatology, 35(12), 837-841 (1996-12-01)
G G Kay et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 29 Suppl 3, 147-150 (1999-08-12)
Although equally potent at blocking the H1 receptor, first- and second-generation antihistamines can be distinguished with respect to their different effects on the central nervous system (CNS). First-generation antihistamines readily cross the blood-brain barrier leading to significant drowsiness, altered mood

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service