Skip to Content
Merck
All Photos(1)

Key Documents

1084000

USP

Butylparaben

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Hydroxybenzoic acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
Beilstein:
1103741
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

parabens

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O(CCCC)C(=O)c1ccc(cc1)O

InChI

1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3

InChI key

QFOHBWFCKVYLES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Butylparaben USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Amcinonide
  • Amcinonide Ointment
  • Amcinonide Cream
  • Loratadine Oral Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darrell R Kapczynski et al.
Vaccine, 33(9), 1197-1205 (2015-01-24)
Vaccination is an important tool in the protection of poultry against avian influenza (AI). For field use, the overwhelming majority of AI vaccines produced are inactivated whole virus formulated into an oil emulsion. However, recombinant vectored vaccines are gaining use
Anchel González-Barriga et al.
PloS one, 10(3), e0121556-e0121556 (2015-03-24)
Myotonic Dystrophy type 1 (DM1) is a multisystemic disease caused by toxic RNA from a DMPK gene carrying an expanded (CTG•CAG)n repeat. Promising strategies for treatment of DM1 patients are currently being tested. These include antisense oligonucleotides and drugs for
Emeran A Mayer et al.
The Journal of clinical investigation, 125(3), 926-938 (2015-02-18)
Tremendous progress has been made in characterizing the bidirectional interactions between the central nervous system, the enteric nervous system, and the gastrointestinal tract. A series of provocative preclinical studies have suggested a prominent role for the gut microbiota in these
L Susta et al.
Veterinary pathology, 52(1), 120-131 (2014-02-11)
To characterize the clinicopathologic features of recently described genotypes of Newcastle disease virus (NDV), 1 representative strain of genotype XIV and 2 of genotype XVII, all isolated from West Africa, were used to infect groups of ten 4-week-old specific pathogen-free
Xiang-Yu Zhang et al.
Biochemical and biophysical research communications, 451(2), 202-207 (2014-08-01)
Fas-associated protein with death domain (FADD) has been implicated in T lymphocytes, but the nature of FADD-dependent mechanism in early T cell development has not been completely elucidated. In this study, using T-cell specific deletion mice, we observed that FADD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service