Skip to Content
Merck
All Photos(1)

Key Documents

1368609

USP

Lisinopril

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-1-[N2-(1-carboxy-3-phenylpropyl)-lysyl-proline dihydrate, MK-521

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31O5N3 · 2H2O
CAS Number:
Molecular Weight:
441.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lisinopril

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H]O[H].[H]O[H].NCCCC[C@H](N[C@@H](CCc1ccccc1)C(O)=O)C(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C21H31N3O5.2H2O/c22-13-5-4-9-16(19(25)24-14-6-10-18(24)21(28)29)23-17(20(26)27)12-11-15-7-2-1-3-8-15;;/h1-3,7-8,16-18,23H,4-6,9-14,22H2,(H,26,27)(H,28,29);2*1H2/t16-,17-,18-;;/m0../s1

InChI key

CZRQXSDBMCMPNJ-ZUIPZQNBSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lisinopril USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Lisinopril and Hydrochlorothiazide Tablets
  • Lisinopril Compounded Oral Suspension
  • Lisinopril Tablets

Biochem/physiol Actions

Angiotensin converting enzyme (ACE) inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J Bright et al.
Australasian journal on ageing, 34(1), 33-37 (2013-10-15)
The Alcohol-Related Problems Survey (ARPS) reliably classifies drinking as non-hazardous, hazardous or harmful using scoring algorithms that consider quantity and frequency of alcohol use alone and in combination with health conditions, medication-use and functional status. Because it has been developed
Lisinopril-induced isolated visceral angioedema: review of ACE-inhibitor-induced small bowel angioedema.
M F Abdelmalek et al.
Digestive diseases and sciences, 42(4), 847-850 (1997-04-01)
K L Goa et al.
Drugs, 52(4), 564-588 (1996-10-01)
Following establishment of its efficacy in hypertension and congestive heart failure, the ACE inhibitor lisinopril has now been shown to reduce mortality and cardiovascular morbidity in patients with myocardial infarction when administered as early treatment. The ability of lisinopril to
Ann Raes et al.
Journal of the renin-angiotensin-aldosterone system : JRAAS, 8(1), 3-12 (2007-05-10)
To investigate the antihypertensive efficacy, dosing, tolerability and effects on growth of lisinopril (off label-use) in paediatric patients during long-term treatment. We conducted a retrospective analysis of data from 123 patients treated with lisinopril in a paediatric nephrology clinic over
K Simpson et al.
Drugs, 59(5), 1149-1167 (2000-06-14)
The ACE inhibitor lisinopril is a lysine derivative of enalaprilat, the active metabolite of enalapril. In patients with heart failure, maximum pharmacodynamic effects are produced 6 to 8 hours after administration of the drug and persist for 12 to 24

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service