Skip to Content
Merck
All Photos(1)

Key Documents

1229001

USP

Droperidol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-[1-[3-(p-Fluorobenzoyl)propyl]-1,2,3,6-tetrahydro-4-pyridyl]-2-benzimidazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22FN3O2
CAS Number:
Molecular Weight:
379.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

droperidol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCC(=CC2)N3C(=O)Nc4ccccc34

InChI

1S/C22H22FN3O2/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28/h1-2,4-5,7-11H,3,6,12-15H2,(H,24,28)

InChI key

RMEDXOLNCUSCGS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Cure et al.
The Cochrane database of systematic reviews, (4)(4), CD002830-CD002830 (2004-10-21)
People suffering from acute psychotic illnesses, especially those associated with agitated or violent behaviour, may require urgent pharmacological tranquillisation or sedation. Droperidol, a butyrophenone neuroleptic, has been used for this purpose in several countries. To estimate the effects of droperidol
Kate McKeage et al.
Drugs, 66(16), 2123-2147 (2006-11-23)
Droperidol (Dehydrobenzperidol, Dehidrobenzoperidol, Dridol, Droleptan, Inapsine) is a dopamine D(2) receptor antagonist that has been widely used in adults and children for the prevention and treatment of postoperative nausea and vomiting (PONV) over several decades and, more recently, for the
R A Chambers et al.
The Journal of clinical psychiatry, 60(10), 664-667 (1999-11-05)
As admission criteria to inpatient units become more focused on patient safety and behavioral instability, primary treatment often requires use of medications that need to be quick, safe, and effective for control of agitation. This article reviews the evidence that
John H Shale et al.
The Journal of clinical psychiatry, 64(5), 500-505 (2003-05-21)
Droperidol had become a standard treatment for sedating severely agitated or violent patients in both psychiatric and medical emergency departments. However, several recent articles have suggested that droperidol may have a quinidine-like effect similar to that of thioridazine in inducing
E S Brown et al.
Clinical neuropharmacology, 21(5), 316-318 (1998-10-28)
There is little information in the literature concerning the use of droperidol in psychiatry. This article presents three cases in which extremely agitated and treatment-refractory persons with mixed mania derived benefit from droperidol administered orally. Symptomatic improvement, including decreased agitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service