Skip to Content
Merck
All Photos(3)

Key Documents

D122

Sigma-Aldrich

Domperidone

powder, ≥98% (HPLC)

Synonym(s):

4-(5-Chloro-2-oxo-1-benzimidazolinyl)-1-[3-(2-oxobenzimidazolinyl)propyl]piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24ClN5O2
CAS Number:
Molecular Weight:
425.91
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: >10 mg/mL
0.1 M HCl: soluble
0.1 M NaOH: slightly soluble
H2O: insoluble

originator

Johnson & Johnson

SMILES string

Clc1ccc2N(C3CCN(CCCN4C(=O)Nc5ccccc45)CC3)C(=O)Nc2c1

InChI

1S/C22H24ClN5O2/c23-15-6-7-20-18(14-15)25-22(30)28(20)16-8-12-26(13-9-16)10-3-11-27-19-5-2-1-4-17(19)24-21(27)29/h1-2,4-7,14,16H,3,8-13H2,(H,24,29)(H,25,30)

InChI key

FGXWKSZFVQUSTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier; anti-emetic.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Domperidone, a specific in vitro dopamine antagonist, devoid of in vivo central dopaminergic activity.
P M Laduron et al.
Biochemical pharmacology, 28(14), 2161-2165 (1979-07-15)
Domperidone.
M C Champion
General pharmacology, 19(4), 499-505 (1988-01-01)
Badriul Hegar et al.
The Turkish journal of pediatrics, 55(1), 1-7 (2013-05-23)
Gastroesophageal reflux (GER) is a normal physiologic process occurring several times per day in healthy infants. Regurgitation is defined as the passage of refluxed gastric contents into the pharynx or mouth, sometimes with expulsion out of the mouth. There are
Savio C Reddymasu et al.
The American journal of gastroenterology, 102(9), 2036-2045 (2007-05-10)
Domperidone is a dopamine-2 receptor antagonist. It acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine. Unlike metoclopramide, it does not cause any adverse
R N Brogden et al.
Drugs, 24(5), 360-400 (1982-11-01)
Domperidone is a dopamine antagonist that does not readily enter the central nervous system. Given parenterally or orally it increases gastric emptying of liquids and increases lower oesophageal sphincter pressure in healthy subjects. The antiemetic and pharmacodynamic profile of domperidone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service