Skip to Content
Merck
All Photos(1)

Key Documents

1162002

USP

Cytarabine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cytosine β-D-arabinofuranoside, (β-D-Arabinofuranosyl)cytosine, Ara-C, Arabinocytidine, Arabinosylcytosine, Cytarabine, Cytosine arabinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O5
CAS Number:
Molecular Weight:
243.22
Beilstein:
89175
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cytarabine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O

InChI

1S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16)/t4-,6-,7+,8-/m1/s1

InChI key

UHDGCWIWMRVCDJ-CCXZUQQUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cytarabine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cytarabine for Injection

Biochem/physiol Actions

Ara-C incorporates into DNA and inhibits DNA replication by forming cleavage complexes with topoisomerase I resulting in DNA fragmentation; does not inhibit RNA synthesis. Anti-leukemia agent.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Yang et al.
Leukemia, 28(6), 1280-1288 (2013-11-26)
Blockade of immune checkpoints is emerging as a new form of anticancer therapy. We studied the expression of programmed death ligand 1 (PD-L1), PD-L2, programmed death 1 (PD-1) and cytotoxic T lymphocyte-associated antigen 4 (CTLA4) mRNA in CD34+ cells from
Fazila Asmar et al.
Oncotarget, 5(7), 1912-1925 (2014-04-12)
MiR34A, B and C have been implicated in lymphomagenesis, but information on their role in normal CD19+ B-cells (PBL-B) and de novo diffuse large B-cell lymphoma (DLBCL) is limited. We show that in normal and activated B-cells miR34A-5p plays a
Xiangyu Ge et al.
Frontiers in pharmacology, 12, 716177-716177 (2021-08-31)
Neuroinflammation plays a crucial role in neurodegenerative diseases such as Alzheimer's disease (AD) and Parkinson's disease (PD), and anti-inflammation has been considered as a potential therapeutic strategy. Achyranthes bidentate polypeptide fraction k (ABPPk) was shown to protect neurons from death
Alona Rudchenko et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(26), 8865-8874 (2014-06-27)
Synapses in autonomic ganglia represent the final output of various CNS structures that regulate the function of the periphery. Normally, these excitatory cholinergic-nicotinic synapses produce large suprathreshold EPSPs on sympathetic and parasympathetic neurons to convey signals from the CNS. However
M R Sapienza et al.
Leukemia, 28(8), 1606-1616 (2014-02-08)
Blastic plasmacytoid dendritic cell neoplasm (BPDCN) is a rare disease of controversial origin recently recognized as a neoplasm deriving from plasmacytoid dendritic cells (pDCs). Nevertheless, it remains an orphan tumor with obscure biology and dismal prognosis. To better understand the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service