Skip to Content
Merck
All Photos(1)

Documents

1140430

USP

Clopidogrel bisulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogensulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClNO2S · H2SO4
CAS Number:
Molecular Weight:
419.90
Beilstein:
9967887
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clopidogrel

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OS(O)(=O)=O.COC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl

InChI

1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/t15-;/m0./s1

InChI key

FDEODCTUSIWGLK-RSAXXLAASA-N

Gene Information

human ... P2RY12(64805)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clopidogrel bisulfate USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Clopidogrel Bisulfate
  • Clopidogrel Tablets
  • Clopidogrel Compounded Oral Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clopidogrel Compounded Oral Suspension
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(5), 1111-1111 (2014)
Clopidogrel Bisulfate
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 38(1), 1109-1109 (2020)
Kailas S Khomane et al.
Journal of pharmaceutical sciences, 101(7), 2408-2416 (2012-04-11)
Being a density violator, clopidogrel bisulfate (CLP) polymorphic system (forms I and II) allows us to study individually the impact of molecular packing (true density) and thermodynamic properties such as heat of fusion on the compaction behavior. These two polymorphs
Clopidogrel Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 33(1), 1111-1111 (2020)
Kyoung-Woo Seo et al.
Clinical therapeutics, 36(11), 1588-1594 (2014-09-15)
Platelet-function suppression with antiplatelet therapy is effective in preventing and treating cardiovascular disease. Clopidogrel is a thienopyridine derivative that blocks platelet activation by adenosine diphosphate receptor binding. This study demonstrates the effects of generic clopidogrel bisulfate in comparison to branded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service