Skip to Content
Merck
All Photos(1)

Key Documents

SML0331

Sigma-Aldrich

Prasugrel

≥98% (HPLC)

Synonym(s):

5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate, CS 747, CS-747, Effient, Efient, LY640315

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20FNO3S
CAS Number:
Molecular Weight:
373.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL (warmed at 60 °C)

storage temp.

2-8°C

SMILES string

CC(=O)Oc1cc2CN(CCc2s1)C(C(=O)C3CC3)c4ccccc4F

InChI

1S/C20H20FNO3S/c1-12(23)25-18-10-14-11-22(9-8-17(14)26-18)19(20(24)13-6-7-13)15-4-2-3-5-16(15)21/h2-5,10,13,19H,6-9,11H2,1H3

InChI key

DTGLZDAWLRGWQN-UHFFFAOYSA-N

Gene Information

human ... P2RY12(64805)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Prasugrel is a platelet inhibitor that reduces the aggregation of platelets by irreversible binding to P2Y12 receptors.
Prasugrel is a platelet inhibitor that reduces the aggregation of platelets by irreversible binding to P2Y12 receptors. Prasugrel interacts in an irreversible manner with the residues Cys97 and Cys175 of the human P2Y12-receptor.
Prasugrel is a thienopyridine prodrug and is considered to be more potent than clopidogrel. It shows a faster generation and also reduces thrombotic events.[1] Prasugrel is less dependent on the CYP (cytochrome P450) enzymes for its conversion to active metabolite.[2]

Features and Benefits

This compound is featured on the P2 Receptors: P2Y G-Protein Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan F Iglesias et al.
Lancet (London, England), 394(10205), 1243-1253 (2019-09-07)
Newer-generation drug-eluting stents that combine ultrathin strut metallic platforms with biodegradable polymers might facilitate vascular healing and improve clinical outcomes in patients with acute myocardial infarction undergoing primary percutaneous coronary intervention (PCI) compared with contemporary thin strut second-generation drug-eluting stents.
Shigeru Saito et al.
Circulation journal : official journal of the Japanese Circulation Society, 78(7), 1684-1692 (2014-04-25)
Prasugrel is an antiplatelet agent that shows more prompt, potent, and consistent platelet inhibition than clopidogrel. The objective of this study was to confirm the efficacy and safety of prasugrel at loading/maintenance doses of 20/3.75 mg. Japanese patients (n=1,363) with
Antonio Tello-Montoliu et al.
Platelets, 27(8), 777-783 (2016-06-04)
Everolimus-eluting bioabsorbable scaffolds (BVSs) have exhibited similar long-term clinical outcomes compared to its everolimus-eluting metallic counterparts. However, reports from earlier studies have shown a signal for an increased rate of stent thrombosis. The aim of the current investigation is to
Matti K Itkonen et al.
Clinical pharmacology and therapeutics, 104(3), 495-504 (2017-11-25)
The oxidation of montelukast is mainly mediated by cytochrome P450 (CYP) 2C8, but other mechanisms may contribute to its disposition. In healthy volunteers, we investigated the effects of two widely used P2Y12 inhibitors on montelukast pharmacokinetics. Clopidogrel (300 mg on day
Jeffrey A Bakal et al.
European heart journal, 36(6), 385-92a-385-92a (2014-07-12)
Several methods provide new insights into understanding clinical trial composite endpoints, using both conventional and novel methods. The TRILOGY ACS trial is used as a contemporary example to prospectively compare these methods side by side. The traditional time-to-first-event, Andersen-Gill recurrent

Articles

Surface receptors for extracellular nucleotides are called P2 receptors. Learn more about P2Y G protein-coupled receptors, GPCRs, and ionotropic P2 receptors (P2X ligand-gated ion channels, LGICs).

Surface receptors for extracellular nucleotides are called P2 receptors. Learn more about P2Y G protein-coupled receptors, GPCRs, and ionotropic P2 receptors (P2X ligand-gated ion channels, LGICs).

Surface receptors for extracellular nucleotides are called P2 receptors. Learn more about P2Y G protein-coupled receptors, GPCRs, and ionotropic P2 receptors (P2X ligand-gated ion channels, LGICs).

Surface receptors for extracellular nucleotides are called P2 receptors. Learn more about P2Y G protein-coupled receptors, GPCRs, and ionotropic P2 receptors (P2X ligand-gated ion channels, LGICs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service