Skip to Content
Merck
All Photos(1)

Documents

1045009

USP

Atropine sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Atropine sulfate salt monohydrate, α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Tropine tropate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H23NO3)2 · H2SO4 · H2O
CAS Number:
Molecular Weight:
694.83
Beilstein:
6109275
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atropine

manufacturer/tradename

USP

mp

189-192 °C (A) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.OS(O)(=O)=O.CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3.CN4[C@H]5CC[C@@H]4C[C@@H](C5)OC(=O)C(CO)c6ccccc6

InChI

1S/2C17H23NO3.H2O4S.H2O/c2*1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12;1-5(2,3)4;/h2*2-6,13-16,19H,7-11H2,1H3;(H2,1,2,3,4);1H2/t2*13-,14+,15+,16?;;

InChI key

JPKKQJKQTPNWTR-CHYDPLAESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atropine sulfate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Atropine Sulfate Injection
  • Atropine Sulfate Ophthalmic Solution
  • Atropine Sulfate Ophthalmic Ointment
  • Diphenoxylate Hydrochloride and Atropine Sulfate Tablets
  • Diphenoxylate Hydrochloride and Atropine Sulfate Oral Solution
  • Atropine

Biochem/physiol Actions

Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miklós Palotai et al.
Neurochemical research, 39(8), 1477-1483 (2014-06-05)
The nociceptin/orphanin FQ peptide (NOP) receptor and its endogenous ligand plays role in several physiologic functions of the central nervous system, including pain, locomotion, anxiety and depression, reward and drug addiction, learning and memory. Previous studies demonstrated that the NOP-receptor
M K Schultz et al.
Neurotoxicology and teratology, 44, 89-104 (2014-06-20)
The progression of epileptiform activity following soman (GD) exposure is characterized by a period of excessive cholinergic activity followed by excessive glutamatergic activity resulting in status epilepticus, which may lead to neuropathological damage and behavioral deficits. Caramiphen edisylate is an
A I Jobling et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(2), 696-710 (2014-11-14)
Age-related macular degeneration (AMD) is a leading cause of vision loss, characterized by drusen deposits and thickened Bruch's membrane (BM). This study details the capacity of nanosecond laser treatment to reduce drusen and thin BM while maintaining retinal structure. Fifty
Francois Malecaze et al.
Investigative ophthalmology & visual science, 55(10), 6712-6721 (2014-09-25)
To characterize the expression of the bone morphogenetic protein-1 (BMP-1)/tolloid-like proteinases (collectively called BTPs), which include BMP-1, mammalian tolloid (mTLD), and mammalian tolloid-like 1 (mTLL-1) and 2 (mTLL-2), as well as the associated proteins procollagen C-proteinase enhancers (PCPE-1 and -2)
Dafne M Silberman et al.
PloS one, 9(6), e98831-e98831 (2014-06-05)
The retina is one of the major energy consuming tissues within the body. In this context, synaptic transmission between light-excited rod and cone photoreceptors and downstream ON-bipolar neurons is a highly demanding energy consuming process. Sirtuin 6 (SIRT6), a NAD-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service