Skip to Content
Merck
All Photos(1)

Key Documents

SU853005

Supelco

Amberlite XAD®-2 polymeric adsorbent

20-60 mesh, pkg of 5 kg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52

Product Name

Amberlite XAD®-2, pkg of 5 kg

Agency

suitable for ASTM® D4861
suitable for EPA Method 10A, Method 23 and OTM-45

Quality Level

form

solid

packaging

pkg of 5 kg

technique(s)

LPLC: suitable
solid phase extraction (SPE): suitable

surface area

~300 m2/g

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

polymer

particle size

20-60 mesh

pore size

~0.65 mL/g pore volume
90 Å mean pore size

density

1.02 g/mL at 25 °C (true wet)(lit.)
1.08 g/mL at 25 °C (skeletal)(lit.)

separation technique

reversed phase

SMILES string

N([C@@H](CCC(=O)OC(C)(C)C)C=O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C24H27NO5/c1-24(2,3)30-22(27)13-12-16(14-26)25-23(28)29-15-21-19-10-6-4-8-17(19)18-9-5-7-11-20(18)21/h4-11,14,16,21H,12-13,15H2,1-3H3,(H,25,28)/t16-/m0/s1

InChI key

BCIPGSZQUDLGSY-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite XAD®-2 is a strongly acidic ion-exchange resin.
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Specifications:

Application

Amberlite XAD®-2 may be used as a stationary phase material for the solid phase extraction (SPE) of phenolic acids and flavonols from honey samples prior to their identification by high performance liquid chromatography-diode array detection (HPLC-DAD), phthalate esters (PAEs) from hydroalcoholic food beverages prior to their determination by gas chromatography (GC) equipped with flame ionization detector (FID) as well as ion-trap mass spectrometry detector (IT/MS), benzoic and sorbic acids in quince jam prior to their analysis by isocratic HPLC combined with photodiode array (PDA) detector and organophosphorus pesticides in natural and drinking waters prior to their determination by GC-FID or GC coupled to nitrogen-phosphorus detector (NPD).
Polyaromatic adsorbent resin for hydrophobic compounds up to MW 20,000: phenols, organic removal, surfactants, aroma compounds, antibiotic recovery. May be used as support for catalyst or metals removal. For cleaned US EPA versions, see Supelpak-2 and Supelpak-2B.
     

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Amberlite is a trademark of DuPont de Nemours, Inc.
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase extraction procedure for determination of phenolic acids and some flavonols in honey.
Michalkiewicz A, et al.
Journal of Chromatography A, 1187(1-2), 18-24 (2008)
Continuous solid-phase extraction and gas chromatographic determination of organophosphorus pesticides in natural and drinking waters.
Ballesteros E and Parrado MJ
Journal of Chromatography A, 1029(1-2), 267-273 (2004)
Simultaneous determination of benzoic and sorbic acids in quince jam by HPLC.
Ferreira IMPLVO, et al.
Food Research International, 33(2), 113-117 (2000)
Study of XAD-2 adsorbent for the enrichment of trace levels of phthalate esters in hydroalcoholic food beverages and analysis by gas chromatography coupled with flame ionization and ion-trap mass spectrometry detectors.
Cinelli G, et al.
Food Chemistry, 146(1-2), 181-187 (2014)
Tim J Causon et al.
Journal of chromatography. A, 1217(21), 3519-3524 (2010-04-21)
Alternative approaches to conventional acetonitrile gradient methods for reversed-phase liquid chromatographic analysis of intact proteins have been investigated using commercial poly(styrene-co-divinylbenzene) monolithic columns (Dionex ProSwift RP-2H and RP-4H). Alternative solvents to acetonitrile (2-propanol and methanol) coupled with elevated temperatures demonstrated

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service