Skip to Content
Merck
All Photos(1)

Key Documents

XAD7

Supelco

Amberlite XAD7HP polymeric adsorbent

20-60 mesh

Synonym(s):

weak anion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

Product Name

Amberlite XAD7HP, 20-60 mesh

vapor density

<1 (vs air)

vapor pressure

17 mmHg ( 20 °C)

form

beads

autoignition temp.

800 °F

technique(s)

LPLC: suitable

surface area

380 m2/g

matrix

acrylic

matrix active group

polymer

particle size

20-60 mesh
560-710 μm

pore size

0.5 mL/g pore volume
300-400 Å mean pore size

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Application

Weakly polar adsorbent resin for compounds up to 60,000 MW: insulin, fulvic and humic compounds, dry waste, organic removal and recovery, and antibiotic recovery.

Other Notes

Moderately polar acrylic resin
Two particle sizes are available:
Mesh 20-60 is Cat. No. XAD7
Mesh 20-50 is Cat. No. 06445

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamad Shazeli Che Zain et al.
Antioxidants (Basel, Switzerland), 9(4) (2020-04-23)
Oil palm (Elaeis guineensis Jacq.) leaves (OPL) are widely available at zero cost in Southeast Asia countries, especially in Malaysia and Indonesia due to large scale oil palm plantations. OPLs contain a large amount of flavonoids in particular flavonoid C-glycosides
Guillermo Schmeda-Hirschmann et al.
Scientific reports, 10(1), 7070-7070 (2020-04-29)
The Chilean hazelnut (Gevuina avellana Mol., Proteaceae) is a southern South American nut consumed as a snack and included in different preparations of traditional Chilean cuisine. Recently we described the fatty acid profile, oxylipins, phenolic compounds, as well as the
Zhibin Liang et al.
mBio, 10(3) (2019-05-30)
Zeamines are a family of polyamino phytotoxins produced by Dickeya zeae EC1. These phytotoxins are also potent antibiotics against a range of microorganisms. To understand how D. zeae EC1 can protect itself from the antimicrobial activity of zeamines, we tested
Johanna Josefine Ostberg-Potthoff et al.
Nutrients, 11(5) (2019-05-30)
An activity-guided search for compounds influencing glucose metabolism in extracts from aronia (Aronia melanocarpa, A.), pomegranate (Punica granatum L., P.), and red grape (Vitis vinifera, RG) was carried out. The three extracts were fractionated by means of membrane chromatography to
Yixiang Liu et al.
Journal of agricultural and food chemistry, 65(15), 3179-3189 (2017-04-01)
Whether all dietary polyphenols nourish the eyes via oral supplementation is controversial. Given that passage of dietary polyphenols across the blood-retina barrier (BRB) is the precondition for polyphenols to exhibit ocular benefits, the BRB permeability of polyphenols was assessed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service