Skip to Content
Merck
All Photos(1)

Key Documents

40353-U

Supelco

48 Component Indoor Air Standard

certified reference material, 1000 μg/mL each component in methanol: water (19:1), ampule of 1 mL

Synonym(s):

Indoor Air Quality Calibration Standard, 48 components

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in methanol: water (19:1)

application(s)

environmental

format

single component solution

storage temp.

-10 to -25°C

Application


  • Monitoring and analysis of indoor air pollutants: The 48 Component Indoor Air Standard is crucial for the accurate monitoring and characterization of volatile organic compounds (VOCs) within various environments, providing essential data for assessing air quality and exposure risks (Gushit et al., 2022).

  • Research on air quality effects on health: This standard aids in the study of the impact of indoor air pollutants on human health, especially in controlled environments where precise quantification of VOCs is necessary to link exposure levels to potential health outcomes (Zwozdziak et al., 2016).

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetone
    Benzene
    Bromodichloromethane
    1-Butanol
    2-Butanone
    Chloroform
    Dibromochloromethane
    Decane
    Decanal
    1,4-Dichlorobenzene
    1,2-Dichloroethane
    Dichloromethane
    1,2-Dichloropropane
    2,4-Dimethylpentane
    Dodecane
    Ethyl alcohol, Pure
    Ethylbenzene
    2-Ethyltoluene
    3-Ethyltoluene
    4-Ethyltoluene
    Heptane
    Hexane
    Hexadecane
    (R)-(+)-Limonene
    4-Methyl-2-pentanone
    Nonanal
    Nonane
    Octane
    Pentadecane
    (1S)-(-)-alpha-Pinene
    (-)-beta-Pinene
    1-Propanol
    2-Propanol
    Styrene
    Tetrachloroethylene
    Tetradecane
    1,2,4,5-Tetramethylbenzene
    Toluene
    Trichloroethylene
    Tridecane
    1,2,3-Trimethylbenzene
    1,2,4-Trimethylbenzene
    1,3,5-Trimethylbenzene
    2,2,4-Trimethylpentane
    Undecane
    o-Xylene
    m-Xylene
    p-Xylene
    See All (48)

Signal Word

Danger

WGK

WGK 3

Flash Point(F)

closed cup

Flash Point(C)

closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service