Skip to Content
Merck
All Photos(3)

Key Documents

57298-U

Supelco

SPME Fiber Assembly

greener alternative

DVB/CAR/PDMS, for use with autosampler, Nitinol-core (NIT), 23 ga, 50 μm (DVB layer), 30 μm (CAR/PDMS layer)

Synonym(s):

DVB/CAR/PDMS SPME Fiber Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

Product Name

SPME fiber assembly 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub

material

StableFlex fiber
plain gray hub

Quality Level

needle size

23 ga

packaging

pk of 3

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

30 μm (CAR/PDMS layer)
50 μm (DVB layer)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

DVB/CAR/PDMS coating

application(s)

food and beverages

compatibility

Autosampler
for analyte group flavors (volatiles and semivolatiles, C3-20) (MW 40-275)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) is used for the analysis of VOC emissions from historic plastic and rubber materials using headspace-solid phase microextraction with gas chromatography coupled with mass spectrometry.

Features and Benefits

Designed for use with Merlin Microseal sealing system; can be used with other septumless systems.

Other Notes

Appearance may vary. Darker spots may be visible on the fiber but will not affect performance.

Footnote

Coating bonded to a flexible fused silica core, yielding a more suitable coating on a less breakable fiber. There may be a slight difference in extraction selectivity compared to the same coating on a standard fused silica core.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The development of a SPME-GC/MS method for the analysis of VOC emissions from historic plastic and rubber materials.
Curran K, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 124, 909-918 (2016)
Sanja Risticevic et al.
Journal of chromatography. A, 1251, 208-218 (2012-07-10)
Metabolomics currently represents one of the fastest growing high-throughput molecular analysis platforms that refer to the simultaneous and unbiased analysis of metabolite pools constituting a particular biological system under investigation. In response to the ever increasing interest in development of
Amparo Gamero et al.
Journal of chromatography. A, 1272, 1-7 (2012-12-19)
MicroVinification platforms are used for screening purposes to study aroma development in wine. These high-throughput methodologies require flavor analysis techniques that allow fast detection of a high number of aroma compounds which often appear in very low concentrations (μg/l). In
Sílvia M Rocha et al.
Journal of chromatography. A, 1252, 155-163 (2012-07-11)
Metabolomics represents an emerging issue that can aid in the diagnosis and/or prognosis of different diseases. Metabolomic study of urine is particularly interesting as it can be on the base of the developing of new faster and non-invasive methodologies. In
M Caldeira et al.
Journal of chromatography. A, 1254, 87-97 (2012-07-28)
Allergic asthma represents an important public health issue, most common in the paediatric population, characterized by airway inflammation that may lead to changes in volatiles secreted via the lungs. Thus, exhaled breath has potential to be a matrix with relevant

Articles

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

Protocols

In this article, SPME-GC was successfully used to identify many volatile compounds present in the headspace over several spirits.

GC Analysis of Terpenes in Cannabis on Equity®-1 after Headspace SPME using 50/30 μm DVB/CAR/PDMS Fiber

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service