Skip to Content
Merck
All Photos(3)

Key Documents

57347-U

Supelco

SPME Fiber Holder

greener alternative

for use with CTC CombiPAL, Gerstel MPS2

Synonym(s):

Solid Phase Microextraction Fiber Holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

solid phase microextraction (SPME): suitable

application(s)

food and beverages

compatibility

for use with CTC CombiPAL, Gerstel MPS2

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

The holder protects the coated fiber, and controls exposure of the fiber during analyte adsorption and desorption. The holder is reusable indefinitely and accepts the replaceable fiber assembly. First time users must order both a holder and a fiber assembly.

Fiber Holder for Manual Sampling

An adjustable depth guide positions the fiber for sampling and for correct placement in the heated zone of the GC injection port. The fiber can be locked in the exposed position.

Fiber Holder for Automated Sampling or HPLC Analysis

Use this fiber holder with a Varian 8100/8200 AutoSampler or with our SPME/HPLC interface. An SPME upgrade kit is necessary for operation with the Varian AutoSampler - contact Varian Instrument Division for information concerning system requirements.

Fiber Holder for CTC CombiPAL/CombiPAL-xt and Gerstel MPS2
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application


  • Surface characterization of commercial fibers for solid-phase microextraction and related problems in their application.: This article provides a comprehensive analysis of the surface properties of various commercial SPME fibers, including those used in SPME Fiber Holders. The study identifies potential issues related to the application of SPME in analytical chemistry, particularly focusing on how these fibers interact with different analytes and solvents. This information is crucial for chemists looking to optimize their SPME techniques for more accurate and reliable sample analysis (Haberhauer-Troyer et al., 2000).



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)
Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)

Articles

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

See All

Protocols

In this article, SPME-GC was successfully used to identify many volatile compounds present in the headspace over several spirits.

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service