Skip to Content
Merck
All Photos(1)

Key Documents

47745-U

Supelco

Styrene

analytical standard

Synonym(s):

Phenylethylene, Vinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CH2
CAS Number:
Molecular Weight:
104.15
Beilstein:
1071236
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.6 (vs air)

vapor pressure

12.4 mmHg ( 37.7 °C)
4.3 mmHg ( 15 °C)

CofA

current certificate can be downloaded

autoignition temp.

914 °F

expl. lim.

6.1 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.546 (lit.)

bp

145-146 °C (lit.)

mp

−31 °C (lit.)

density

0.906 g/mL at 25 °C

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Styrene is a volatile compound, used in the manufacture of numerous types of plastic, glass fiber-reinforced resins, protective coatings, synthetic rubber and ion exchange resins.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Styrene may be used as an analytical reference for the quantification of the analyte in water samples using gas chromatography coupled to mass spectrometry.
Styrene may be used as an analytical reference standard for the quantification of the analyte in air samples using thermal desorption-gas chromatography-mass spectrometry technique and comestible oil samples using headspace-mass spectrometry/gas chromatography coupled to mass spectrometry.

Other Notes

Product contains 4-tert-butylcatechol as stabilizer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Headspace?Mass spectrometry with alternative chromatographic separation using a column switching system for the screening and determination of BTEX and Styrene in comestible oil
Toledo C, et al.
Analytical Letters, 43(5), 843-850 (2010)
Development of a thermal desorption-gas chromatography-mass spectrometry method for determination of styrene in air. Application to workplace air
Martin V-F V, et al.
Analusis, 28(8), 737-742 (2000)
Solid-phase microextraction method for the quantitative analysis of styrene in water.
Silva FC, et al.
Journal of Chromatographic Science, 38(7), 315-318 (2000)
A comprehensive evaluation of the potential health risks associated with occupational and environmental exposure to styrene.
Joshua T Cohen et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 5(1-2), 1-265 (2002-05-16)
Daniela Saviello et al.
Analytica chimica acta, 843, 59-72 (2014-08-26)
Synchrotron-based Fourier transform infrared micro-spectroscopy (SR-μFTIR) was used to map photo-oxidative degradation of acrylonitrile-butadiene-styrene (ABS) and to investigate the presence and the migration of additives in historical samples from important Italian design objects. High resolution (3×3 μm(2)) molecular maps were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service