Skip to Content
Merck
All Photos(1)

Key Documents

45993

Supelco

Styrene

analytical standard

Synonym(s):

Phenylethylene, Vinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CH2
CAS Number:
Molecular Weight:
104.15
Beilstein:
1071236
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.6 (vs air)

vapor pressure

12.4 mmHg ( 37.7 °C)
4.3 mmHg ( 15 °C)

autoignition temp.

914 °F

quality

stabilized

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.1 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.546 (lit.)

bp

145-146 °C (lit.)

mp

−31 °C (lit.)

density

0.906 g/mL at 25 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
petroleum

format

neat

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Styrene has been used as an analytical standard for the analysis of volatile metabolic profile along in vitro differentiation of human induced pluripotent stem cells using gas chromatography coupled to mass spectrometry (GC-MS).[1]

Caution

limited shelf life

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

GC/MS-based analysis of volatile metabolic profile along in vitro differentiation of human induced pluripotent stem cells
Capuano R, et al.
Scientific reports (2017)
Bum Gun Kwon et al.
Environmental pollution (Barking, Essex : 1987), 188, 45-49 (2014-02-21)
Beach sand and seawater taken from the coastlines of the North-East Pacific Ocean and Hawaii State were investigated to determine the causes of global chemical contamination from polystyrene (PS). All samples were found to contain styrene monomer (SM), styrene dimers (SD)
Pavel Vodicka et al.
Mutation research, 511(3), 239-254 (2002-06-29)
Styrene is an important industrial chemical that has shown genotoxicity in many toxicology assays. This is believed to be related to the DNA-binding properties of styrene-7,8-oxide (SO), a major metabolite of styrene. In this review, we have summarized knowledge on
Niall D O'Leary et al.
FEMS microbiology reviews, 26(4), 403-417 (2002-11-05)
The last few decades have seen a steady increase in the global production and utilisation of the alkenylbenzene, styrene. The compound is of major importance in the petrochemical and polymer-processing industries, which can contribute to the pollution of natural resources
James J Collins et al.
Epidemiology (Cambridge, Mass.), 24(2), 195-203 (2013-01-25)
Epidemiologic studies have reported increased risk of lymphohematopoietic cancers, lung cancer, and pancreatic cancer after exposure to styrene, although findings across studies are not consistent. We update a large study of reinforced plastic industry workers with relatively high exposures to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service