Skip to Content
Merck
All Photos(1)

Key Documents

40212

Supelco

Bromoform solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBr3
CAS Number:
Molecular Weight:
252.73
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 601
suitable for EPA 601

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

BrC(Br)Br

InChI

1S/CHBr3/c2-1(3)4/h1H

InChI key

DIKBFYAXUHHXCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromoform can be used as a solvent in palynological preparation. It is the largest form of available atmospheric organic bromine.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Air?sea flux of bromoform: Controls, rates, and implications
Quack B and Wallace.R.W.D
Global Biogeochemical Cycles, 17 (2003)
Sodium polytungstate, a new non-toxic alternative to bromoform in heavy liquid separation
Munsterman D and Kerstholt S
Review of Palaeobotany and Palynology, 91, 417-422 (1996)
Bichismita Sahu et al.
The Journal of organic chemistry, 74(6), 2601-2604 (2009-02-26)
Addition of bromoform to conjugated nitroalkenes in the presence of Mg provided beta-tribromomethyl nitroalkanes in good to excellent yields and diastereoselectivity. These novel Michael adducts, formed under radical conditions, underwent elimination of HBr in the same pot under reflux to
Lisa George et al.
The Journal of chemical physics, 135(12), 124503-124503 (2011-10-07)
Iso-polyhalomethanes are known reactive intermediates that play a pivotal role in the photochemistry of halomethanes in condensed phases. In this work, iso-bromoform (iso-CHBr(3)) and its deuterated isotopomer were characterized by matrix isolation infrared and UV/visible spectroscopy, supported by ab initio
Szilvia Pothoczki et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 22(40), 404211-404211 (2011-03-10)
New neutron and x-ray diffraction measurements are reported on liquid chloroform, CHCl(3), and bromoform, CHBr(3). Experimental total scattering structure factors have been interpreted by the reverse Monte Carlo method of structural modelling. Partial radial distribution functions, intramolecular bond angle distributions

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service