Skip to Content
Merck
All Photos(1)

Key Documents

36972

Supelco

Bromoform

amylene stabilized, analytical standard

Synonym(s):

Tribromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBr3
CAS Number:
Molecular Weight:
252.73
Beilstein:
1731048
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

8.7 (vs air)

vapor pressure

5 mmHg ( 20 °C)

quality

amylene stabilized

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.595 (lit.)

bp

146-150 °C (lit.)

mp

5-8 °C (lit.)

density

2.89 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

BrC(Br)Br

InChI

1S/CHBr3/c2-1(3)4/h1H

InChI key

DIKBFYAXUHHXCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

106.3 °F - closed cup - (own results)

Flash Point(C)

41.3 °C - closed cup - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacey L Carrier et al.
The journal of physical chemistry. A, 114(3), 1548-1555 (2009-12-09)
Ultrafast photolysis of bromoform (CHBr(3)) with a 267 nm pulse of light followed by broadband transient electronic absorption identifies the photoproducts and follows their evolution in both neat bromoform and cyclohexane solutions. In neat bromoform, a species absorbing at 390
David R Geter et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 43(9), 1405-1412 (2005-06-01)
Folate and folic acid are forms of the B vitamin that are involved in the synthesis, repair, and functioning of DNA and are required for the production and maintenance of cells. Low levels of folate have been associated with several
S K Pal et al.
The Journal of chemical physics, 138(12), 124501-124501 (2013-04-06)
Ultrafast deep-ultraviolet through near infrared (210-950 nm) transient absorption spectroscopy complemented by ab initio multiconfigurational calculations offers a global description of the photochemical reaction pathways of bromoform following 255-nm excitation in methylcyclohexane and acetonitrile solutions. Photoexcitation of CHBr3 leads to
Carolina Lourencetti et al.
Environment international, 45, 59-67 (2012-05-11)
This first study of trihalomethanes (THMs) in swimming pools using bromine agents for water disinfection under real conditions shows that the mixtures of these compounds are largely dominated by bromoform in a similar process as chloroform becomes the dominant THM
Szilvia Pothoczki et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 22(40), 404211-404211 (2011-03-10)
New neutron and x-ray diffraction measurements are reported on liquid chloroform, CHCl(3), and bromoform, CHBr(3). Experimental total scattering structure factors have been interpreted by the reverse Monte Carlo method of structural modelling. Partial radial distribution functions, intramolecular bond angle distributions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service