Skip to Content
Merck
All Photos(1)

Documents

40147-U

Supelco

C8-C40 Alkanes Calibration Standard

in dichloromethane (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CofA

current certificate can be downloaded

Quality Level

packaging

pkg of 1 mL

concentration

in dichloromethane (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Qualitative standard. May require warming before use.

Analyte

Description

    Octane 500 μg/mL
    Nonane 500 μg/mL
    Decane 500 μg/mL
    Undecane 500 μg/mL
    Dodecane 500 μg/mL
    Tridecane 500 μg/mL
    Tetradecane 500 μg/mL
    Pentadecane 500 μg/mL
    Hexadecane 500 μg/mL
    Heptadecane 500 μg/mL
    Octadecane 500 μg/mL
    Nonadecane 500 μg/mL
    Pristane 500 μg/mL
    Eicosane 500 μg/mL
    Phytane 500 μg/mL
    Docosane 500 μg/mL
    Tricosane 500 μg/mL
    Tetracosane 500 μg/mL
    Pentacosane 500 μg/mL
    Heneicosane 500 μg/mL
    Hexacosane 500 μg/mL
    Heptacosane 500 μg/mL
    Octacosane 500 μg/mL
    Nonacosane 500 μg/mL
    Triacontane 500 μg/mL
    Hentriacontane 500 μg/mL
    Dotriacontane 500 μg/mL
    Tritriacontane 500 μg/mL
    Tetratriacontane 500 μg/mL
    Pentatriacontane 500 μg/mL
    Hexatriacontane 500 μg/mL
    Heptatriacontane 500 μg/mL
    Octatriacontane 500 μg/mL
    Nonatriacontane
    Tetracontane
    Carbon disulfide 5000 μg/mL
    See All (36)

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Nie et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 59, 152803-152803 (2019-04-22)
Chrysanthemi Flos (CF), as a popular traditional Chinese medicine (TCM), has five main cultivars in China, namely "Chuju", "Boju", "Gongju", "Huaiju" and "Hangju". Due to their habitats and processing methods, great quality variations occur yet no systematical study has ever
Rafael Mascoloti Spréa et al.
Food & function, 11(2), 1292-1303 (2020-01-31)
Aromatic plants have been used since antiquity as seasoning ingredients to impart unique flavours to foods, and also as folk medicines, and are currently used as a source of several compounds of interest for different industries. Lovage (Levisticum officinale W.J.D.
Sarika Gupta et al.
Environmental monitoring and assessment, 189(1), 43-43 (2017-01-01)
Delhi is one of the most polluted cities in the world. The generation of aerosols in the lower atmosphere of the city is mainly due to a large amount of natural dust advection and sizable anthropogenic activities. The compositions of
M M Uribe-Flores et al.
Journal of applied microbiology, 127(2), 495-507 (2019-05-12)
The aim of this study was to investigate the dynamic changes in the bacterial structure and potential interactions of an acclimatized marine microbial community during a light crude oil degradation experiment. The bacterial community effectively removed 76·49% of total petroleum
Zechariah C Harris et al.
Journal of insect physiology, 120, 103991-103991 (2019-12-11)
This study investigated the water balance physiology of Venezillo arizonicus, a land isopod endemic to the Southwest Desert Ecoregion of North America. Evaporative water losses were measured in two ways: gravimetric in dry air, and by perfusing animals with dry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service