Skip to Content
Merck
All Photos(1)

Key Documents

40084

Supelco

Toluene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chun-Xia Zhao et al.
Methods in molecular biology (Clifton, N.J.), 996, 179-194 (2013-03-19)
The self-organization of peptide-based nanostructures at a confined fluid-fluid interface, for example, the air-water or oil-water interface, is important in the context of stabilizing macroscopic soft-matter foams and emulsions. The unique ability to design interfacial nanostructures by controlling the subtle
Yevgeny Tendler et al.
International journal of molecular medicine, 31(3), 540-546 (2013-01-23)
The tumor-suppressor gene p53 encodes a phosphoprotein involved in the control of cell growth. p53 expression and function have been documented in malignancy, apoptosis and the aging processes. Recently, p53 has been mapped and characterized in the normal cornea across
Patrick R Burney et al.
The journal of physical chemistry. B, 117(9), 2662-2670 (2013-02-08)
Ionic liquids (ILs) and organic chemicals can be used as solvents in biochemical reactions to influence the structural and dynamic features of the enzyme, sometimes detrimentally. In this work we report the results for molecular dynamics simulations of Candida rugosa
Yajie Qian et al.
Chemosphere, 91(5), 717-723 (2013-03-08)
Due to the large diameter and small surface, the contaminant degradation by conventional calcium peroxide (CaO2) is slow with high dosage required. The aggregation of conventional CaO2 also makes it difficult to operate. Nanoscale CaO2 was therefore synthesized and applied
N A Dorofeeva et al.
Zhurnal evoliutsionnoi biokhimii i fiziologii, 49(2), 137-143 (2013-06-26)
In the present work we analyzed a possibility of interaction of protein p53, family members of the ERK1/2 signaling cascade, and the transcription factor CREB in regulation of functional activity of dopaminergic neurons. There were considered neurons of Substantia nigra

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service