Skip to Content
Merck
All Photos(1)

Documents

25461

Supelco

Carboxen®-1006 PLOT Capillary GC Column

L × I.D. 30 m × 0.53 mm, average thickness 30 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710

material

fused silica

description

PLOT = Porous Layer Open Tubular

manufacturer/tradename

Carboxen®

parameter

≤25-250 °C temperature (isothermal or programmed)

average thickness

30 μm

df

30 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

30 m × 0.53 mm

matrix active group

Carbon molecular sieve phase

application(s)

chemicals and industrial polymers
food and beverages

column type

capillary PLOT

Looking for similar products? Visit Product Comparison Guide

General description

Carboxen® Adsorbent is ideally used for solid-phase microextraction (SPME) and does efficient extraction of small polar molecules from aqueous solution. Porous-layer open-tubular (PLOT) columns are a powerful tool used for analyzing fixed gases, light hydrocarbons and volatile solvents in gas chromatography method. They are mostly preferred over traditional packed columns.
Application: This column is ideal for the separation of many permanent gas components (such as helium, hydrogen, nitrogen, carbon monoxide, methane, and carbon dioxide), and light hydrocarbons (C2-C3) in the same analysis. It is ideal for resolving formaldehyde/water/methanol (formalin) mixtures and monitoring impurities in ethylene. This column can be used with high flow rates and rapid temperature programs to ensure excellent, fast separations.
USP Code: None
Phase:
  • Carbon molecular sieve
Temp. Limits:
  • Subambient to 250 °C (isothermal or programmed)

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Applications of Solid Phase Microextraction
Pawliszyn J.
Applications of Solid Phase Microextraction, 64-65 (1999)
Porous layer open-tubular capillary columns: preparations, applications and future directions.
Zhenghua Ji
Journal of Chromatography A, 842 (1), 115-142 (1999)
B D Page et al.
Journal of chromatography. A, 873(1), 79-94 (2000-04-11)
The headspace solid-phase microextraction (HS-SPME) efficiencies from vegetable oil of the recently available Carboxen-poly(dimethylsiloxane) (PDMS) and divinylbenzene-Carboxen-PDMS fibres were found to be much greater than those of the PDMS fibre for a number of volatile contaminants. Using these Carboxen-based fibres
Dawn M Chapman et al.
Journal of agricultural and food chemistry, 52(17), 5431-5435 (2004-08-19)
A rapid and automated solid phase microextraction (SPME) stable isotope dilution gas chromatography/mass spectrometry (GC-MS) method for 2-methoxy-3-isobutylpyrazine (MIBP) quantification in red wine was developed. Wines with 30% (w/v) NaCl and 2-methoxy-(2)H(3)-3-isobutylpyrazine internal standard were sampled with a 2 cm
Lucia Giordano et al.
Journal of chromatography. A, 1017(1-2), 141-149 (2003-10-31)
A new method was developed for the determination of 2-furfural (2-F) and 5-methylfurfural (5-MF), two products of Maillard reaction in vinegar, with head-space solid-phase microextraction (HS-SPME) coupled to gas chromatography-mass spectrometry (GC-MS). A divinylbenzene (DVB)/carboxen (CAR)/polydimethylsiloxane (PDMS) fibre was used

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Related Content

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service