Skip to Content
Merck
All Photos(1)

Key Documents

18913-1AMP

Supelco

F.A.M.E. Mix, C20:1-C20:5 Unsaturates

certified reference material, ampule of 50 mg (Neat)

Synonym(s):

Fatty Acid Methyl Ester Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 50 mg (Neat)

concentration

(approx. 20% (by weight) each component)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

carboxylic acid
ester

shipped in

dry ice

storage temp.

−20°C

Application

It was used in analysing fatty acid methyl esters using GC method, in a study which was performed to understand manipulation of oil synthesis in Nannochloropsis strain NIES-2145 with a phosphorus starvation–inducible promoter from Chlamydomonas reinhardtii.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    cis-11,14-Eicosadienoic acid methyl ester ~ 10 mg
    cis-11-Eicosenoic acid methyl ester ~ 10 mg
    cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester ~ 10 mg
    cis-5,8,11,14-Eicosatetraenoic acid methyl ester ~ 10 mg
    cis-11,14,17-Eicosatrienoic acid methyl ester ~ 10 mg

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masako Iwai et al.
Frontiers in microbiology, 6, 912-912 (2015-10-07)
Microalgae accumulate triacylglycerols (TAGs) under conditions of nutrient stress. Phosphorus (P) starvation induces the accumulation of TAGs, and the cells under P starvation maintain growth through photosynthesis. We recently reported that P starvation-dependent overexpression of type-2 diacylglycerol acyl-CoA acyltransferase (CrDGTT4)
Emma Teuling et al.
Journal of agricultural and food chemistry, 65(36), 7989-8002 (2017-07-14)
Photosynthetic unicellular organisms are considered as promising alternative protein sources. The aim of this study is to understand the extent to which these green sources differ with respect to their gross composition and how these differences affect the final protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service