Skip to Content
Merck
All Photos(1)

Key Documents

10190-U

Supelco

Carbosieve Carbon Adsorbent

matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g

Synonym(s):

polymeric adsorbent, DVB, 80-100 mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
SB.54

product name

Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g

product line

Carbosieve

Quality Level

form

powder or granules

packaging

bottle of 10 g

technique(s)

LPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

surface area

~1059 m2/g

matrix

Carbosieve S-II

matrix active group

carbon

particle size

80-100 mesh

pore size

~0.01 cm3/g mesoporosity
~0.45 cm3/g microporosity
~0 cm3/g macroporosity
~6-15 Å pore diameter

density

~0.61 g/mL (free fall density)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

A carbon molecular sieve (CMS) is the porous carbon skeletal framework that remains after pyrolysis of a polymeric precursor. These particles are:
  • Spherical (better packed bed performance than granular particles)
  • Hard and non-friable (pack well, will not break)
  • Highly porous (high surface areas)
  • Used for molecules with an analyte size relative to C2-C5 n-alkanes
  • Hydrophobic (can be used in high humidity environments)

Generally, CMS adsorbents offer greater relative adsorptive strength compared to spherical graphitized polymer carbon (SGPC) and graphitized carbon black (GCB) adsorbents. Our Carbosieve products are a type of CMS adsorbent.
  • Have non-tapered pores
  • Very strong adsorptive strength due to only containing micropores
  • Provide great performance for many small, volatile analytes that most adsorbents have trouble retaining

For more information about any of our specialty carbon adsorbents, please visit sigma-aldrich.com/carbon

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water adsorption capacity of the solid adsorbents Tenax TA, Tenax GR, Carbotrap, Carbotrap C, Carbosieve SIII, and Carboxen 569 and water management techniques for the atmospheric sampling of volatile organic trace gases.
Detlev H et al.
Analytical Chemistry, 67 (23), 4380-4386 (1995)
Yannick Juillet et al.
The Analyst, 130(6), 977-982 (2005-05-25)
Among the chemicals belonging to the schedules of the Chemical Weapons Convention (CWC), sampling and analysis of highly volatile compounds such as hydrogen cyanide (HCN) require special consideration. The latter is present in numerous old chemical weapons that are stockpiled
C Y Peng et al.
Journal of environmental monitoring : JEM, 2(4), 313-324 (2001-03-16)
Air sampling, using sorbents, thermal desorption and gas chromatography, is a versatile method for identifying and quantifying trace levels of volatile organic compounds (VOCs). Thermal desorption can provide high sensitivity, appropropriate choices of sorbents and method parameters can accommodate a
Mohammad Reza Samarghandi et al.
Journal of research in health sciences, 14(3), 227-232 (2014-09-12)
Toluene is a volatile organic compound, one of 189 hazardous air pollutants (HAPs) and the most important pollutant found in most industries and indoor environments; owing to its adverse health, toluene must be treated before being released into the environment.
John L Martin et al.
Journal of occupational and environmental hygiene, 1(6), 363-370 (2004-07-09)
In an effort to assess the scope of occupational exposures to ethylene, the Olefins Panel of the American Chemistry Council designed and conducted a research project to develop and apply a sampling and analytical method to measure workplace exposure. The

Articles

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service