Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbosieve Carbon Adsorbent
EC Number:
231-153-3
Products
Product Category
sieves (5)
Brand
Supelco (5)
Physical Form
Product Line
Separation Technique
Quality Segment
Package Size
Pore Size (Å)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
10184
Carbosieve Carbon Adsorbent,
matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Expand
View Pricing
10198
Carbosieve Carbon Adsorbent,
matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Expand
View Pricing
10189
Carbosieve Carbon Adsorbent,
matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Expand
View Pricing
10190-U
Carbosieve Carbon Adsorbent,
matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g
Expand
View Pricing
10199
Carbosieve Carbon Adsorbent,
matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Expand
View Pricing