Skip to Content
Merck
All Photos(1)

Key Documents

Z742663

Molecular Grinding Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.74

manufacturer/tradename

G-BioSciences 786-138

packaging

1 ea of

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Includes:
Molecular Grinding Resin 5 x 0.5 ml resin
Molecular Grinding Resin has been specifically developed for effectively grinding biological samples for extraction of DNA, RNA and proteins. The resin is high tensile micro-particles, which effectively disrupt nuclei and other cell organelles. Molecular Grinding Resin does not bind protein or nucleic acids. Simply mix the biological sample with Molecular Grinding Resin and grind or homogenize the sample. Unlike sand or other grinding materials, Molecular Grinding Resin is easy to work with and does not damage genomic DNA or high molecular mass proteins. We routinely use Molecular Grinding Resin for isolation of 100kb genomic DNA and RNA. Molecular Grinding Resin can be used with any homogenization technique, including high speed mechanical grinding such as Polytron and high-energy sonicators. Molecular Grinding Resin is also available with disposable matching pestles and tubes.

Legal Information

Molecular Grinding Resin is a trademark of Geno Technology, Inc.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service