Skip to Content
Merck
All Photos(1)

Key Documents

Z3003

Sigma-Aldrich

Zardaverine

powder, ≥98% (HPLC)

Synonym(s):

6-(4-Difluoromethoxy-3-methoxyphenyl)-3(2H)-pyridazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F2N2O3
CAS Number:
Molecular Weight:
268.22
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Zardaverine,

Quality Level

SMILES string

COc1cc(ccc1OC(F)F)C2=NNC(=O)C=C2

InChI

1S/C12H10F2N2O3/c1-18-10-6-7(2-4-9(10)19-12(13)14)8-3-5-11(17)16-15-8/h2-6,12H,1H3,(H,16,17)

InChI key

HJMQDJPMQIHLPB-UHFFFAOYSA-N

Application

Zardaverine has been used as a phosphodiesterase inhibitor:
  • to study its effects on human colonic epithelial cells
  • to study its effects on junctional proteins in Sertoli cells
  • to study its effects on phosphodiesterase 3 A (PDE3A) expression in various tumor cells

Biochem/physiol Actions

Zardaverine is a derivative of pyridazinone. It shows anti-tumor and anti-proliferative activity in varied cell lines. Zardaverine possesses broncho-dilatory activity and exhibits therapeutic effects against asthma.
Selective inhibitor of phosphodiesterase III/IV (PDE3/4).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Fischer et al.
Biochemical pharmacology, 45(12), 2399-2404 (1993-06-22)
Phosphodiesterase inhibitors were used as a tool to manipulate cellular nucleotide levels in vitro and in vivo. The lipopolysaccharide (LPS)-induced release of tumor necrosis factor alpha (TNF-alpha) from mouse peritoneal macrophages was inhibited by prostaglandin E2 with an IC50 of
D T Schmidt et al.
British journal of pharmacology, 131(8), 1607-1618 (2001-01-05)
Non-selective inhibitors of cyclic nucleotide phosphodiesterase (PDE) block allergen-induced contraction of passively sensitized human airways in vitro by a dual mechanism involving a direct relaxant effect on smooth muscle and inhibition of histamine and cysteinyl leukotriene (LT) release from airways.
A Hatzelmann et al.
British journal of pharmacology, 114(4), 821-831 (1995-02-01)
1. The effect of non-selective (3-isobutyl-1-methylxanthine, IBMX; theophylline) and type IV- or type III/IV-selective (rolipram, RP 73401; zardaverine, tolafentrine) phosphodiesterase (PDE) inhibitors on human eosinophil functions was investigated. 2. For this purpose human eosinophils were purified from blood of healthy
K F Rabe et al.
The American journal of physiology, 266(5 Pt 1), L536-L543 (1994-05-01)
The effects of the nonselective phosphodiesterase (PDE) inhibitor 3-isobutyl-1-methylxanthine (IBMX) and the selective PDE inhibitors motapizone (type III), rolipram (type IV), zardaverine (type III/IV), and zaprinast (type V and I) on prostaglandin F2 alpha (PFG2 alpha)-induced tone in human pulmonary
K F Rabe et al.
The American journal of physiology, 264(5 Pt 1), L458-L464 (1993-05-01)
The effects of the nonselective phosphodiesterase (PDE)-inhibitor 3-isobutyl-1-methylxanthine (IBMX) and the selective PDE inhibitors SKF 94120 (type III), rolipram (type IV), zardaverine (type III/IV), and zaprinast (type V) on inherent tone in human airways were investigated. Substantial relaxation was achieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service