Skip to Content
Merck
All Photos(1)

Key Documents

Y0626

Sigma-Aldrich

Yeast Nitrogen Base Without Amino Acids

Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements

Synonym(s):

ynb without amino acids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.85

grade

Molecular Biology
for molecular biology

Quality Level

sterility

non-sterile

form

powder

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

pH

5.2-5.6(0.67% solution)

application(s)

food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Candida spp.
nonselective for Pichia spp.
nonselective for Saccharomyces spp.
nonselective for Zygosaccharomyces spp.
yeasts

General description

Yeast Nitrogen Base is a highly-referenced growth medium used for the cultivation of yeast. This nutrient-rich microbial broth contains nitrogen, vitamins, trace elements and salts.
Yeast Nitrogen Base is considered ideal for culturing wild type yeast. The presence of amino acid is known to prevent selectable marker utilization. Hence, this media is formulated without amino acids and is useful for cloning and manipulation of yeast artificial chromosome.

Application

Suitable for use in classifying yeasts based on amino acid and carbon requirements. Addition of amino acids and carbon source (usually glucose) may be required.
Yeast Nitrogen Base Without Amino Acids has been used:
  • in synthetic complete supplement mixture (SC)-Leu media used for Y. lipolytica culturing and characterization
  • as a component of selective liquid media for uracil auxothrophic growth in yeast
  • to evaluate the transformation efficiencies of S. cerevisiae strains, under different nutrient conditions

Components

Component, amount (μg/L unless indicated)

Nitrogen Sources:
Ammonium sulfate, 5.0 g/L

Vitamins:
Biotin, 2.0
Calcium pantothenate, 400
Folic acid, 2.0
Inositol, 2.0 mg/L
Nicotinic acid, 400
p-Aminobenzoic acid, 200
Pyridoxine HCl, 400
Riboflavin, 200
Thiamine HCL, 400

Trace Elements:
Boric acid, 500
Copper sulfate, 40
Potassium iodide, 100
Ferric chloride, 200
Manganese sulfate, 400
Sodium molybdate, 200
Zinc sulfate, 400

Salts:
Potassium phosphate monobasic, 1.0 g/L
Magnesium sulfate, 0.5 g/L
Sodium chloride, 0.1 g/L
Calcium chloride, 0.1 g/L

Preparation Note

1. Prepare a 10× stock solution by suspending 6.7 g of yeast nitrogen base, 5 g of glucose or an equivalent amount of other carbohydrate, and 5-10 mg of the desired amino acid in 100 ml of distilled water.
2. Warm if necessary to solubilize and sterilize by filtration.
Store 10× stock solution at 2-8 °C. To use, dilute 1:10 with sterile distilled water under aspetic conditions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Daniel Gietz et al.
Nature protocols, 2(1), 38-41 (2007-04-03)
Here, we describe a Library screen transformation protocol using the lithium acetate/single-stranded carrier DNA/PEG method of transformation for Saccharomyces cerevisiae. This method is suitable for screening complex plasmid libraries such as those used for yeast two-hybrid analysis. This procedure takes
T Fernandes et al.
Journal of applied microbiology, 127(3), 845-855 (2019-06-19)
The aim was to isolate, identify and characterize yeasts present in rumen fluid and to select strains showing potential as probiotics. Rumen fluid was sampled from 4 herds of dairy and beef cattle and 77 yeast isolates were identified. Initial
YAC Protocols, 11-11 (2006)
Activating and elucidating metabolism of complex sugars in Yarrowia lipolytica
Ryu S, et al.
Applied and Environmental Microbiology, 82(4), 1334-1345 (2016)
Simultaneous saccharification and fermentation of cellulose in ionic liquid for efficient production of alpha-ketoglutaric acid by Yarrowia lipolytica
Ryu S, et al.
Applied Microbiology and Biotechnology, 99(10), 4237-4244 (2015)

Articles

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Related Content

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service