Skip to Content
Merck
All Photos(1)

Key Documents

XNAB2R

Sigma-Aldrich

Extract-N-Amp Blood PCR Kit

sufficient for 1000 extractions, sufficient for 1000 amplifications

Synonym(s):

Blood direct PCR kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 1000 amplifications
sufficient for 1000 extractions
sufficient for 1000 reactions

Quality Level

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

colorless

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Extract-N-Amp Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by direct PCR. This novel extraction system eliminates the need for any type of purification, organic extraction, centrifugation, heating, filtration or alcohol precipitation. The kit also includes a PCR ReadyMix, specially formulated for amplification directly from the extract. This formulation uses an antibody based hot start, for specific amplification.

Application

Extract-N-Amp Blood PCR Kit has been used to extract DNA from genomes and dried blood spot samples. It has also been used for polymerase chain reaction (PCR).

Features and Benefits

  • Efficient 8 minute prep allows greater speed and throughput
  • No need for any type of purification, organic extraction, centrifugation or alcohol precipitation
  • Simple, 3 step procedure with no special equipment required
  • Hot start antibody included for highly specific PCR amplification of genomic DNA
  • Compatible with multiple format (single tube or 96-well plate)
  • Can be used with whole blood or blood cards
  • Extract stable at 4 °C for at least 6 months

Principle

Genomic DNA is extracted from 10 μl of whole blood by simply adding the Extraction Solution and incubating for 5 minutes at room temperature. The Neutralization Solution is added to the extract to counteract inhibitory substances prior to PCR. A portion of the DNA extract is then added to the specially formulated PCR ReadyMix.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • L3289Lysis Solution for BloodSDS

  • N9784Neutralization Solution for BloodSDS

  • P8115Extract-N-Amp PCR ReadyMix for BloodSDS

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas Wong et al.
BioTechniques, 45(4), 423-424 (2008-10-16)
Sodium bisulfite treatment followed by PCR and DNA sequencing is widely considered the gold standard for the analysis of DNA methylation patterns. However, this technique generally requires substantial quantities of genomic DNA as starting material and is often associated with
Jun Shi et al.
Cancer research, 67(13), 6417-6424 (2007-07-10)
Idiopathic myelofibrosis (IM) is likely the consequence of both the acquisition of genetic mutations and epigenetic changes that silence critical genes that control cell proliferation, differentiation, and apoptosis. We have explored the effects of the sequential treatment with the DNA
Mads V Hollegaard et al.
BMC genomics, 10, 297-297 (2009-07-07)
Identification of disease susceptible genes requires access to DNA from numerous well-characterised subjects. Archived residual dried blood spot samples from national newborn screening programs may provide DNA from entire populations and medical registries the corresponding clinical information. The amount of
Mads Vilhelm Hollegaard et al.
Electrophoresis, 30(14), 2532-2535 (2009-07-30)
Stored surplus of dried blood spot (DBS) samples from neonatal screening programs constitute a vast potential for large genetic epidemiological studies. However, age of the samples and the small amounts of DNA available may limit their usage. In this study
Sandrine Romand et al.
Biotechnology and bioengineering, 113(5), 1094-1101 (2015-11-03)
Chinese Hamster Ovary (CHO) cells are widely used for the large-scale production of recombinant biopharmaceuticals. However, attempts to express IGF-1 (a mutated human Insulin-like growth factor 1 Ea peptide (hIGF-1Ea mut)) in CHO cells resulted in poor cell growth and

Articles

The Extract-N-Amp Blood PCR Kits contain all the reagents needed to rapidly extract and amplify human genomic DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

The Extract-N-Amp Blood PCR Kits contain all the reagents needed to rapidly extract and amplify human genomic DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

The Extract-N-Amp Blood PCR Kits contain all the reagents needed to rapidly extract and amplify human genomic DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

The Extract-N-Amp Blood PCR Kits contain all the reagents needed to rapidly extract and amplify human genomic DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service