Skip to Content
Merck
All Photos(8)

Key Documents

WH0010875M1

Sigma-Aldrich

Monoclonal Anti-FGL2 antibody produced in mouse

clone 6D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-T49, Anti-fibrinogen-like 2, Anti-pT49

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D9, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGL2(10875)

General description

Fibrinogen-like protein 2 (FGL2) belongs to the fibrinogen-like protein family. The FGL2 gene is located on the human chromosome at 7q11.23.

Immunogen

FGL2 (NP_006673, 24 a.a. ~ 123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPG

Application

Monoclonal Anti-FGL2 antibody produced in mouse has been used in immunohistochemistry.

Biochem/physiol Actions

Fibrinogen-like protein 2 (FGL2) exhibits prothrombinase activity. It also shows immune regulatory activity during allograft rejection, abortion, and viral infections. This protein acts as an immune coagulant that produces thrombin directly. FGL2 through the clotting-dependent pathway may play a role in tumor angiogenesis and metastasis. It is involved in the pathogenesis of T helper type 1 (Th1) cytokine-induced fetal loss syndrome and viral-induced fulminant hepatitis. FGL2 gene is involved in modulating regulatory T cells (Treg) function. This protein regulates innate and adaptive immunity. FGL2 may serve as a therapeutic agent for glioblastoma (GBM) by promoting GBM progression.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Shao et al.
Fundamental & clinical pharmacology, 28(1), 42-52 (2012-09-19)
Atorvastatin is not only an antilipemic but also used as an anti-inflammatory medicine in heart disease. Our working hypothesis was that atorvastatin preconditioning could improve the forward blood flow in the no-reflow rats associated with inflammation. We investigated that two
Kai Su et al.
World journal of gastroenterology, 14(39), 5980-5989 (2008-10-22)
To examine the role of Fibrinogen-like protein 2 (fgl2)/fibroleukin in tumor development. Fgl2 has been reported to play a vital role in the pathogenesis in MHV-3 (mouse hepatitis virus) induced fulminant and severe hepatitis, spontaneous abortion, allo- and xeno- graft
Camie W Y Chan et al.
Journal of immunology (Baltimore, Md. : 1950), 170(8), 4036-4044 (2003-04-12)
Fibrinogen-like protein 2 (fgl2)/fibroleukin is a member of the fibrinogen-related protein superfamily. In addition to its established role in triggering thrombosis, it is known to be secreted by T cells. The soluble fgl2 ((s)fgl2) protein generated in a baculovirus expression
Khatri Latha et al.
Journal of the National Cancer Institute, 111(3), 292-300 (2018-06-28)
Virtually all low-grade gliomas (LGGs) will progress to high-grade gliomas (HGGs), including glioblastoma, the most common malignant primary brain tumor in adults. A key regulator of immunosuppression, fibrinogen-like protein 2 (FGL2), may play an important role in the malignant transformation
Liang Shao et al.
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis, 19(1), 19-28 (2012-03-06)
No-reflow phenomenon due to cardiac microvascular dysfunction or disturbance aggravates clinic outcomes of a portion of patients with acute myocardial infarction undergoing percutaneous coronary intervention or thrombolytic therapy. Our working hypothesis was that cardiac microthrombosis would play an important role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service