Skip to Content
Merck
All Photos(3)

Documents

WH0008878M1

Sigma-Aldrich

Monoclonal Anti-SQSTM1 antibody produced in mouse

clone 2C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PDB3, Anti-ZIP3, Anti-p60, Anti-p62, Anti-sequestosome 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SQSTM1(8878)

Immunogen

SQSTM1 (AAH03139.1, 1 a.a. ~ 440 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MASLTVKAYLLGKEDAAREIRRFSFCCSPEPEAEAEAAAGPGPCERLLSRVAALFPALRPGGFQAHYRDEDGDLVAFSSDEELTMAMSYVKDDIFRIYIKEKKECRRDHRPPCAQEAPRNMVHPNVICDGCNGPVVGTRYKCSVCPDYDLCSVCEGKGLHRGHTKLAFPSPFGHLSEGFSHSRWLRKVKHGHFGWPGWEMGPPGNWSPRPPRAGEARPGPTAESASGPSEDPSVNFLKNVGESVAAALSPLGIEVDIDVEHGGKRSRLTPVSPESSSTEEKSSSQPSSCCSDPSKPGGNVEGATQSLAEQMRKIALESEGRPEEQMESDNCSGGDDDWTHLSSKEVDPSTGELQSLQMPESEGPSSLDPSQEGPTGLKEAALYPHLPPEADPRLIESLSQMLSMGFSDEGGWLTRLLQTKNYDIGAALDTIQYSKHPPPL

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabina Berezowska et al.
Methods in molecular biology (Clifton, N.J.), 1560, 189-194 (2017-02-06)
Autophagy is a highly conserved cellular mechanism of "self digestion," ensuring cellular homeostasis, and playing a role in many diseases including cancer. As a stress response mechanism, it may also be involved in cellular response to therapy.LC3 and Sequestosome 1
Angelo Pascarella et al.
Journal of cellular physiology, 233(8), 5829-5837 (2017-12-08)
Autosomal recessive Pompe disease is a lysosomal disorder caused by mutations of the acid-α-glucosidase (GAA) gene. Deficiency of GAA enzyme leads to glycogen accumulation and autophagy impairment in cardiac and skeletal muscles, but also in lymphocytes. Since an effective therapy
Hilke Brühl et al.
Arthritis and rheumatism, 60(5), 1352-1361 (2009-05-01)
Activation of basophils contributes to memory immune responses and results in exacerbation of collagen-induced arthritis (CIA). We undertook the present study to analyze the production and biologic effects of interleukin-3 (IL-3), a strong activator of basophils, in CIA. Arthritis was
Ragini Vuppugalla et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(12), 1446-1454 (2004-09-24)
Nitric oxide (NO) is implicated as a mediator in the decreased catalytic activities of cytochrome P450 (P450) enzymes during inflammation or infection. Here, we examined the time course and the reversibility of the NO effect on P450s using isolated perfused
Olivia Adams et al.
Oncotarget, 7(26), 39241-39255 (2016-10-27)
Esophageal adenocarcinomas (EAC) are aggressive tumors with considerable rates of chemoresistance. Autophagy is a lysosome-dependent degradation process, characterized by the formation of vesicles called autophagosomes, and has been implicated in cancer. Protein light chain 3 B (LC3B) and p62 are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service